Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34177937-34178164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf94
Entrez Gene ID
84970
Gene Name
chromosome 1 open reading frame 94
Gene Aliases
-
UniGene
Hs.194610
Gene Chromosome Location
Chr.1: 34166883-34219131 on Build GRCh38
Transcripts
NM_001134734.1 NM_032884.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4631585 c.267C>A p.L89L Chr.1: 34178056-34178056 on Build GRCh38
1584493 c.261G>T p.E87D Chr.1: 34178050-34178050 on Build GRCh38
5985692 c.314A>G p.Q105R Chr.1: 34178103-34178103 on Build GRCh38
1584494 c.253T>C p.S85P Chr.1: 34178042-34178042 on Build GRCh38
4969705 c.215_216delGA p.R72fs*6 Chr.1: 34178004-34178005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745335397 Chr.1: 34178141-34178141 on Build GRCh38
rs910107980 Chr.1: 34177959-34177959 on Build GRCh38
rs961315964 Chr.1: 34178095-34178095 on Build GRCh38
rs778313456 Chr.1: 34178097-34178097 on Build GRCh38
rs532358753 Chr.1: 34178010-34178010 on Build GRCh38
rs540897178 Chr.1: 34178025-34178025 on Build GRCh38
rs980553476 Chr.1: 34178122-34178122 on Build GRCh38
rs370350958 Chr.1: 34178014-34178014 on Build GRCh38
rs530078092 Chr.1: 34178018-34178018 on Build GRCh38
rs201119842 Chr.1: 34177964-34177964 on Build GRCh38
rs950253120 Chr.1: 34177980-34177980 on Build GRCh38
rs564391103 Chr.1: 34177963-34177963 on Build GRCh38
rs1027430120 Chr.1: 34178116-34178117 on Build GRCh38
rs559485545 Chr.1: 34178139-34178139 on Build GRCh38
rs895970541 Chr.1: 34177978-34177978 on Build GRCh38
rs756417359 Chr.1: 34178022-34178022 on Build GRCh38
rs749794102 Chr.1: 34178108-34178108 on Build GRCh38
rs970101073 Chr.1: 34178121-34178121 on Build GRCh38
rs905974713 Chr.1: 34178069-34178069 on Build GRCh38
rs1054371679 Chr.1: 34178098-34178098 on Build GRCh38
rs940254429 Chr.1: 34177962-34177962 on Build GRCh38
rs546693784 Chr.1: 34178130-34178130 on Build GRCh38
rs771654093 Chr.1: 34178142-34178142 on Build GRCh38
rs890922107 Chr.1: 34177988-34177988 on Build GRCh38
rs1015458963 Chr.1: 34178080-34178080 on Build GRCh38

View Full Product Details