Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 130060741-130061014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAP1
Entrez Gene ID
50807
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1
Gene Aliases
AMAP1, CENTB4, DDEF1, PAG2, PAP, ZG14P
UniGene
Hs.655552
Gene Chromosome Location
Chr.8: 130052104-130443848 on Build GRCh38
Transcripts
NM_001247996.1 NM_018482.3 XM_005250925.1 XM_006716563.3 XM_006716564.1 XM_006716565.3 XM_006716566.1 XM_011517052.2 XM_011517053.1 XM_017013467.1 XM_017013468.1 XM_017013469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3778903 c.2766G>A p.Q922Q Chr.8: 130061005-130061005 on Build GRCh38
3951284 c.2987G>A p.G996E Chr.8: 130060784-130060784 on Build GRCh38
6472355 c.2919C>A p.P973P Chr.8: 130060852-130060852 on Build GRCh38
5710851 c.2842C>T p.P948S Chr.8: 130060929-130060929 on Build GRCh38
6488368 c.2967G>A p.L989L Chr.8: 130060804-130060804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746318209 Chr.8: 130060801-130060801 on Build GRCh38
rs200760463 Chr.8: 130060762-130060762 on Build GRCh38
rs756690941 Chr.8: 130060884-130060884 on Build GRCh38
rs200764058 Chr.8: 130060904-130060904 on Build GRCh38
rs760278832 Chr.8: 130060827-130060827 on Build GRCh38
rs761590323 Chr.8: 130060765-130060765 on Build GRCh38
rs750552166 Chr.8: 130060961-130060961 on Build GRCh38
rs878960677 Chr.8: 130060848-130060848 on Build GRCh38
rs766446321 Chr.8: 130060776-130060776 on Build GRCh38
rs760227312 Chr.8: 130060774-130060774 on Build GRCh38
rs751723274 Chr.8: 130060870-130060870 on Build GRCh38
rs556009108 Chr.8: 130060895-130060895 on Build GRCh38
rs926280708 Chr.8: 130060931-130060931 on Build GRCh38
rs143261202 Chr.8: 130060803-130060803 on Build GRCh38
rs200682369 Chr.8: 130060909-130060909 on Build GRCh38
rs878889766 Chr.8: 130060856-130060856 on Build GRCh38
rs775979019 Chr.8: 130060808-130060808 on Build GRCh38
rs754736965 Chr.8: 130060780-130060780 on Build GRCh38
rs553616348 Chr.8: 130060979-130060979 on Build GRCh38
rs753773187 Chr.8: 130060777-130060777 on Build GRCh38
rs765251399 Chr.8: 130060917-130060917 on Build GRCh38
rs763201843 Chr.8: 130060809-130060809 on Build GRCh38
rs775241259 Chr.8: 130060921-130060921 on Build GRCh38
rs781314525 Chr.8: 130060882-130060882 on Build GRCh38
rs751349147 Chr.8: 130060925-130060925 on Build GRCh38
rs757530775 Chr.8: 130060932-130060932 on Build GRCh38
rs879169122 Chr.8: 130060864-130060864 on Build GRCh38
rs765064792 Chr.8: 130060854-130060854 on Build GRCh38
rs758150325 Chr.8: 130060858-130060858 on Build GRCh38
rs149161756 Chr.8: 130060800-130060800 on Build GRCh38
rs777941033 Chr.8: 130060796-130060796 on Build GRCh38
rs752577734 Chr.8: 130060855-130060855 on Build GRCh38
rs767610218 Chr.8: 130060954-130060954 on Build GRCh38
rs765923132 Chr.8: 130060833-130060833 on Build GRCh38
rs770948515 Chr.8: 130060799-130060799 on Build GRCh38
rs757332068 Chr.8: 130060881-130060881 on Build GRCh38
rs776255255 Chr.8: 130060907-130060907 on Build GRCh38
rs780390053 Chr.8: 130060889-130060889 on Build GRCh38
rs759577453 Chr.8: 130060853-130060853 on Build GRCh38
rs753286556 Chr.8: 130060847-130060847 on Build GRCh38
rs758696528 Chr.8: 130060792-130060792 on Build GRCh38
rs749350012 Chr.8: 130060815-130060815 on Build GRCh38
rs747121611 Chr.8: 130060798-130060798 on Build GRCh38
rs764321312 Chr.8: 130060860-130060860 on Build GRCh38
rs769089064 Chr.8: 130060900-130060900 on Build GRCh38
rs749998843 Chr.8: 130060766-130060766 on Build GRCh38
rs191674894 Chr.8: 130060788-130060788 on Build GRCh38
rs201380090 Chr.8: 130060838-130060838 on Build GRCh38
rs141392919 Chr.8: 130060903-130060903 on Build GRCh38
rs771887908 Chr.8: 130060813-130060813 on Build GRCh38
rs745919113 Chr.8: 130060883-130060883 on Build GRCh38

View Full Product Details