Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30944988-30945213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAP2
Entrez Gene ID
55803
Gene Name
ArfGAP with dual PH domains 2
Gene Aliases
CENTA2, HSA272195, cent-b
UniGene
Hs.514063
Gene Chromosome Location
Chr.17: 30921736-30959193 on Build GRCh38
Transcripts
NM_018404.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7274051 c.596A>G p.Q199R Chr.17: 30944992-30944992 on Build GRCh38
1381998 c.594G>T p.L198L Chr.17: 30944990-30944990 on Build GRCh38
3515802 c.654G>A p.G218G Chr.17: 30945050-30945050 on Build GRCh38
3515801 c.613G>A p.D205N Chr.17: 30945009-30945009 on Build GRCh38
6324927 c.613G>T p.D205Y Chr.17: 30945009-30945009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774137094 Chr.17: 30945096-30945096 on Build GRCh38
rs1017172408 Chr.17: 30945025-30945025 on Build GRCh38
rs373046073 Chr.17: 30945090-30945090 on Build GRCh38
rs921567861 Chr.17: 30945102-30945102 on Build GRCh38
rs559740822 Chr.17: 30945169-30945171 on Build GRCh38
rs376715701 Chr.17: 30945041-30945041 on Build GRCh38
rs983000914 Chr.17: 30945021-30945021 on Build GRCh38
rs188507848 Chr.17: 30945087-30945087 on Build GRCh38
rs987587161 Chr.17: 30945111-30945111 on Build GRCh38
rs755413282 Chr.17: 30945053-30945053 on Build GRCh38
rs773284563 Chr.17: 30945036-30945036 on Build GRCh38
rs780947629 Chr.17: 30945075-30945075 on Build GRCh38
rs369475618 Chr.17: 30945080-30945080 on Build GRCh38
rs772303540 Chr.17: 30945105-30945105 on Build GRCh38
rs751317855 Chr.17: 30945061-30945061 on Build GRCh38
rs759793655 Chr.17: 30945119-30945119 on Build GRCh38
rs765517621 Chr.17: 30945054-30945054 on Build GRCh38
rs754279150 Chr.17: 30945047-30945047 on Build GRCh38
rs933841735 Chr.17: 30945120-30945120 on Build GRCh38
rs745405672 Chr.17: 30945079-30945079 on Build GRCh38
rs370889542 Chr.17: 30945091-30945091 on Build GRCh38
rs760632564 Chr.17: 30945040-30945040 on Build GRCh38
rs756988121 Chr.17: 30945065-30945065 on Build GRCh38

View Full Product Details