Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13012896-13013163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP192
Entrez Gene ID
55125
Gene Name
centrosomal protein 192
Gene Aliases
PPP1R62
UniGene
Hs.100914
Gene Chromosome Location
Chr.18: 12991307-13125053 on Build GRCh38
Transcripts
NM_032142.3 XM_005258107.3 XM_005258109.3 XM_006722326.3 XM_006722327.3 XM_006722330.3 XM_011525673.2 XM_011525675.2 XM_017025803.1 XM_017025804.1 XM_017025806.1 XM_017025807.1

SNP Detail

SNP ID Chromosome Location
rs980498364 Chr.18: 13013044-13013044 on Build GRCh38
rs538516598 Chr.18: 13012981-13012981 on Build GRCh38
rs575374472 Chr.18: 13013101-13013102 on Build GRCh38
rs926860800 Chr.18: 13013039-13013039 on Build GRCh38
rs753980265 Chr.18: 13013066-13013066 on Build GRCh38
rs962850240 Chr.18: 13013020-13013020 on Build GRCh38
rs1025535476 Chr.18: 13012968-13012968 on Build GRCh38
rs766482838 Chr.18: 13013065-13013065 on Build GRCh38
rs925052698 Chr.18: 13013089-13013089 on Build GRCh38
rs901512454 Chr.18: 13012939-13012939 on Build GRCh38
rs950470511 Chr.18: 13012967-13012968 on Build GRCh38
rs755711855 Chr.18: 13012985-13012985 on Build GRCh38
rs969252663 Chr.18: 13013012-13013012 on Build GRCh38
rs149453534 Chr.18: 13013109-13013112 on Build GRCh38
rs750088067 Chr.18: 13012976-13012976 on Build GRCh38
rs372310306 Chr.18: 13013040-13013040 on Build GRCh38
rs567783897 Chr.18: 13012980-13012980 on Build GRCh38
rs1026499522 Chr.18: 13012964-13012964 on Build GRCh38
rs755282491 Chr.18: 13013071-13013071 on Build GRCh38
rs762087792 Chr.18: 13012947-13012947 on Build GRCh38
rs957664148 Chr.18: 13013096-13013096 on Build GRCh38
rs981024833 Chr.18: 13013031-13013031 on Build GRCh38

View Full Product Details