Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42077736-42078001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM109B
Entrez Gene ID
150368
Gene Name
family with sequence similarity 109 member B
Gene Aliases
IPIP27B, Ses2
UniGene
Hs.368312
Gene Chromosome Location
Chr.22: 42074244-42079438 on Build GRCh38
Transcripts
NM_001002034.2 XM_005261373.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4502861 c.624C>T p.L208L Chr.22: 42077917-42077917 on Build GRCh38
217242 c.688T>A p.W230R Chr.22: 42077981-42077981 on Build GRCh38
3708303 c.521G>A p.C174Y Chr.22: 42077814-42077814 on Build GRCh38
6658074 c.564G>A p.A188A Chr.22: 42077857-42077857 on Build GRCh38
5494224 c.706G>A p.V236M Chr.22: 42077999-42077999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776453299 Chr.22: 42077763-42077763 on Build GRCh38
rs1807493 Chr.22: 42077856-42077856 on Build GRCh38
rs200769228 Chr.22: 42077982-42077982 on Build GRCh38
rs866861612 Chr.22: 42077926-42077926 on Build GRCh38
rs761903869 Chr.22: 42077866-42077866 on Build GRCh38
rs778256121 Chr.22: 42077909-42077909 on Build GRCh38
rs945990281 Chr.22: 42077843-42077843 on Build GRCh38
rs781710333 Chr.22: 42077832-42077832 on Build GRCh38
rs753187509 Chr.22: 42077904-42077904 on Build GRCh38
rs754157518 Chr.22: 42077921-42077921 on Build GRCh38
rs761511969 Chr.22: 42077949-42077950 on Build GRCh38
rs574460714 Chr.22: 42077797-42077797 on Build GRCh38
rs957669502 Chr.22: 42077787-42077787 on Build GRCh38
rs763235462 Chr.22: 42077915-42077915 on Build GRCh38
rs747670639 Chr.22: 42077963-42077963 on Build GRCh38
rs768097561 Chr.22: 42077860-42077860 on Build GRCh38
rs756554620 Chr.22: 42077905-42077905 on Build GRCh38
rs766517929 Chr.22: 42077794-42077794 on Build GRCh38
rs147466995 Chr.22: 42077793-42077793 on Build GRCh38
rs751779767 Chr.22: 42077890-42077890 on Build GRCh38
rs897692809 Chr.22: 42077889-42077889 on Build GRCh38
rs774110141 Chr.22: 42077843-42077846 on Build GRCh38
rs866739275 Chr.22: 42077833-42077833 on Build GRCh38
rs531351772 Chr.22: 42077941-42077941 on Build GRCh38
rs762873012 Chr.22: 42077880-42077880 on Build GRCh38
rs746475341 Chr.22: 42077857-42077857 on Build GRCh38
rs766575469 Chr.22: 42077883-42077883 on Build GRCh38
rs768534352 Chr.22: 42077796-42077797 on Build GRCh38
rs373452762 Chr.22: 42077929-42077929 on Build GRCh38
rs926212794 Chr.22: 42077818-42077818 on Build GRCh38
rs764049141 Chr.22: 42077760-42077760 on Build GRCh38
rs769818439 Chr.22: 42077965-42077965 on Build GRCh38
rs886862703 Chr.22: 42077950-42077950 on Build GRCh38
rs762820267 Chr.22: 42077972-42077972 on Build GRCh38
rs375284046 Chr.22: 42077789-42077789 on Build GRCh38
rs193003847 Chr.22: 42077923-42077923 on Build GRCh38
rs200117018 Chr.22: 42077879-42077879 on Build GRCh38
rs778166307 Chr.22: 42077850-42077850 on Build GRCh38
rs139250450 Chr.22: 42077836-42077836 on Build GRCh38
rs937646955 Chr.22: 42077871-42077871 on Build GRCh38
rs770673118 Chr.22: 42077975-42077975 on Build GRCh38
rs755034482 Chr.22: 42077806-42077806 on Build GRCh38
rs769869639 Chr.22: 42077878-42077878 on Build GRCh38
rs756464681 Chr.22: 42077849-42077849 on Build GRCh38
rs767148828 Chr.22: 42077964-42077965 on Build GRCh38
rs746677003 Chr.22: 42077934-42077934 on Build GRCh38
rs1040751995 Chr.22: 42077981-42077981 on Build GRCh38
rs913335538 Chr.22: 42077803-42077803 on Build GRCh38
rs767559219 Chr.22: 42077897-42077897 on Build GRCh38
rs144073756 Chr.22: 42077770-42077770 on Build GRCh38
rs780512734 Chr.22: 42077946-42077946 on Build GRCh38
rs369577093 Chr.22: 42077862-42077862 on Build GRCh38
rs1056349078 Chr.22: 42077881-42077881 on Build GRCh38
rs759630860 Chr.22: 42077894-42077894 on Build GRCh38
rs760105157 Chr.22: 42077756-42077756 on Build GRCh38
rs772709389 Chr.22: 42077966-42077967 on Build GRCh38
rs765194365 Chr.22: 42077786-42077786 on Build GRCh38
rs1007816879 Chr.22: 42077969-42077969 on Build GRCh38
rs868417126 Chr.22: 42077759-42077759 on Build GRCh38

View Full Product Details