Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47223381-47223636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK16
Entrez Gene ID
5127
Gene Name
cyclin dependent kinase 16
Gene Aliases
PCTAIRE, PCTAIRE1, PCTGAIRE, PCTK1
UniGene
Hs.496068
Gene Chromosome Location
Chr.X: 47217881-47229997 on Build GRCh38
Transcripts
NM_001170460.1 NM_006201.4 NM_033018.3 XM_011543920.2 XM_011543921.1 XM_011543922.1 XM_011543923.1 XM_011543924.1 XM_011543925.2 XM_011543926.1 XM_011543927.2 XM_011543928.1 XM_017029569.1 XM_017029570.1 XM_017029571.1 XM_017029572.1 XM_017029573.1

SNP Detail

SNP ID Chromosome Location
rs750593584 Chr.X: 47223583-47223583 on Build GRCh38
rs369198498 Chr.X: 47223515-47223515 on Build GRCh38
rs752824712 Chr.X: 47223584-47223584 on Build GRCh38
rs910017016 Chr.X: 47223565-47223565 on Build GRCh38
rs766632657 Chr.X: 47223596-47223596 on Build GRCh38
rs965731769 Chr.X: 47223476-47223477 on Build GRCh38
rs773807417 Chr.X: 47223525-47223525 on Build GRCh38
rs889705808 Chr.X: 47223480-47223480 on Build GRCh38
rs764501733 Chr.X: 47223554-47223554 on Build GRCh38
rs998247681 Chr.X: 47223419-47223419 on Build GRCh38
rs1050817924 Chr.X: 47223465-47223465 on Build GRCh38
rs759556075 Chr.X: 47223494-47223494 on Build GRCh38
rs994038845 Chr.X: 47223541-47223541 on Build GRCh38
rs1006772902 Chr.X: 47223508-47223508 on Build GRCh38
rs762446019 Chr.X: 47223566-47223566 on Build GRCh38
rs180754755 Chr.X: 47223582-47223582 on Build GRCh38
rs754825902 Chr.X: 47223609-47223609 on Build GRCh38
rs867927569 Chr.X: 47223594-47223594 on Build GRCh38
rs377023226 Chr.X: 47223511-47223511 on Build GRCh38
rs761130276 Chr.X: 47223536-47223536 on Build GRCh38
rs187232602 Chr.X: 47223613-47223613 on Build GRCh38
rs751693661 Chr.X: 47223604-47223604 on Build GRCh38

View Full Product Details