Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50601832-50602105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65C
Entrez Gene ID
140876
Gene Name
family with sequence similarity 65 member C
Gene Aliases
C20orf175, C20orf176
UniGene
Hs.372578
Gene Chromosome Location
Chr.20: 50586108-50692004 on Build GRCh38
Transcripts
NM_001290268.1 NM_080829.3 NR_110890.1 XM_005260294.3 XM_006723713.3 XM_011528578.2 XM_011528579.2 XM_011528580.2 XM_011528581.2 XM_011528584.2 XM_011528585.2 XM_011528586.2 XM_017027681.1 XM_017027682.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6426142 c.1619A>G p.Q540R Chr.20: 50602100-50602100 on Build GRCh38
6201463 c.1626C>T p.L542L Chr.20: 50602093-50602093 on Build GRCh38
7251521 c.1633C>T p.R545W Chr.20: 50602086-50602086 on Build GRCh38
6384959 c.1634G>A p.R545Q Chr.20: 50602085-50602085 on Build GRCh38
1412427 c.1642C>T p.L548L Chr.20: 50602077-50602077 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs917188923 Chr.20: 50602059-50602059 on Build GRCh38
rs73272554 Chr.20: 50601863-50601863 on Build GRCh38
rs1023575140 Chr.20: 50601944-50601944 on Build GRCh38
rs900713457 Chr.20: 50601966-50601966 on Build GRCh38
rs760271998 Chr.20: 50602068-50602068 on Build GRCh38
rs577488890 Chr.20: 50602086-50602086 on Build GRCh38
rs775249600 Chr.20: 50602065-50602065 on Build GRCh38
rs768025692 Chr.20: 50602071-50602071 on Build GRCh38
rs780379658 Chr.20: 50602049-50602049 on Build GRCh38
rs761448115 Chr.20: 50602085-50602085 on Build GRCh38
rs566907555 Chr.20: 50601981-50601981 on Build GRCh38
rs373678317 Chr.20: 50602050-50602050 on Build GRCh38
rs975349726 Chr.20: 50602045-50602045 on Build GRCh38
rs548799196 Chr.20: 50601950-50601950 on Build GRCh38
rs927137340 Chr.20: 50601886-50601886 on Build GRCh38
rs894041455 Chr.20: 50601906-50601906 on Build GRCh38
rs760026090 Chr.20: 50602044-50602049 on Build GRCh38
rs535587278 Chr.20: 50601945-50601945 on Build GRCh38
rs894001197 Chr.20: 50602075-50602075 on Build GRCh38
rs776399232 Chr.20: 50602079-50602079 on Build GRCh38
rs1011101381 Chr.20: 50602080-50602080 on Build GRCh38
rs1035492403 Chr.20: 50601968-50601968 on Build GRCh38
rs1050859309 Chr.20: 50601856-50601856 on Build GRCh38
rs937147480 Chr.20: 50601901-50601901 on Build GRCh38
rs909749052 Chr.20: 50602022-50602022 on Build GRCh38
rs568143686 Chr.20: 50601907-50601907 on Build GRCh38
rs189577349 Chr.20: 50601898-50601898 on Build GRCh38
rs772155963 Chr.20: 50602063-50602063 on Build GRCh38
rs988512381 Chr.20: 50601989-50601989 on Build GRCh38
rs774295149 Chr.20: 50602059-50602059 on Build GRCh38
rs748305817 Chr.20: 50602055-50602055 on Build GRCh38
rs770917317 Chr.20: 50602058-50602058 on Build GRCh38
rs951501620 Chr.20: 50601976-50601976 on Build GRCh38
rs181586857 Chr.20: 50601904-50601904 on Build GRCh38
rs949423848 Chr.20: 50601915-50601915 on Build GRCh38
rs775759564 Chr.20: 50601960-50601960 on Build GRCh38
rs962615097 Chr.20: 50602042-50602042 on Build GRCh38
rs558951276 Chr.20: 50602053-50602053 on Build GRCh38
rs1000078272 Chr.20: 50601870-50601870 on Build GRCh38
rs781526412 Chr.20: 50602054-50602054 on Build GRCh38
rs751019874 Chr.20: 50601867-50601868 on Build GRCh38
rs1048165868 Chr.20: 50601943-50601943 on Build GRCh38

View Full Product Details