Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 790222-790451 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHTF18
Entrez Gene ID
63922
Gene Name
chromosome transmission fidelity factor 18
Gene Aliases
C16orf41, C321D2.2, C321D2.3, C321D2.4, CHL12, Ctf18, RUVBL
UniGene
Hs.153850
Gene Chromosome Location
Chr.16: 788622-798074 on Build GRCh38
Transcripts
NM_022092.2 XM_005255470.1 XM_005255471.3 XM_011522572.1 XM_017023532.1 XM_017023533.1 XM_017023534.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6145295 c.683A>T p.K228M Chr.16: 790253-790253 on Build GRCh38
6145291 c.737C>G p.S246* Chr.16: 790384-790384 on Build GRCh38
4579327 c.700C>T p.R234W Chr.16: 790347-790347 on Build GRCh38
4504832 c.675C>T p.S225S Chr.16: 790245-790245 on Build GRCh38
2151792 c.1298A>G p.E433G Chr.16: 790354-790354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375307452 Chr.16: 790428-790428 on Build GRCh38
rs371408062 Chr.16: 790391-790391 on Build GRCh38
rs749523674 Chr.16: 790374-790374 on Build GRCh38
rs555115888 Chr.16: 790420-790420 on Build GRCh38
rs772010365 Chr.16: 790384-790384 on Build GRCh38
rs199680337 Chr.16: 790270-790270 on Build GRCh38
rs753543725 Chr.16: 790354-790354 on Build GRCh38
rs757808554 Chr.16: 790362-790362 on Build GRCh38
rs370733302 Chr.16: 790327-790327 on Build GRCh38
rs201930845 Chr.16: 790280-790280 on Build GRCh38
rs370280399 Chr.16: 790350-790350 on Build GRCh38
rs201124441 Chr.16: 790309-790309 on Build GRCh38
rs772541637 Chr.16: 790281-790281 on Build GRCh38
rs554395065 Chr.16: 790244-790244 on Build GRCh38
rs909292170 Chr.16: 790432-790432 on Build GRCh38
rs201557926 Chr.16: 790351-790351 on Build GRCh38
rs372399848 Chr.16: 790260-790260 on Build GRCh38
rs137893865 Chr.16: 790294-790294 on Build GRCh38
rs200430188 Chr.16: 790368-790368 on Build GRCh38
rs763214391 Chr.16: 790333-790335 on Build GRCh38
rs775896514 Chr.16: 790334-790334 on Build GRCh38
rs766009475 Chr.16: 790353-790353 on Build GRCh38
rs745911940 Chr.16: 790370-790370 on Build GRCh38
rs373047501 Chr.16: 790246-790246 on Build GRCh38
rs200098278 Chr.16: 790328-790328 on Build GRCh38
rs759240881 Chr.16: 790251-790251 on Build GRCh38
rs3765264 Chr.16: 790409-790409 on Build GRCh38
rs772096807 Chr.16: 790242-790242 on Build GRCh38
rs983907644 Chr.16: 790323-790323 on Build GRCh38
rs547904005 Chr.16: 790344-790344 on Build GRCh38
rs3765262 Chr.16: 790307-790307 on Build GRCh38
rs755230515 Chr.16: 790269-790269 on Build GRCh38
rs543803181 Chr.16: 790292-790292 on Build GRCh38
rs539357230 Chr.16: 790404-790404 on Build GRCh38
rs376304766 Chr.16: 790320-790320 on Build GRCh38
rs1033301992 Chr.16: 790272-790272 on Build GRCh38
rs769676263 Chr.16: 790302-790302 on Build GRCh38
rs761169566 Chr.16: 790311-790311 on Build GRCh38
rs773087466 Chr.16: 790386-790386 on Build GRCh38
rs540964444 Chr.16: 790306-790306 on Build GRCh38
rs539283811 Chr.16: 790415-790415 on Build GRCh38
rs572535954 Chr.16: 790264-790264 on Build GRCh38
rs759222673 Chr.16: 790395-790395 on Build GRCh38
rs758707757 Chr.16: 790356-790356 on Build GRCh38
rs779039722 Chr.16: 790419-790419 on Build GRCh38
rs756330363 Chr.16: 790371-790371 on Build GRCh38
rs759813876 Chr.16: 790296-790296 on Build GRCh38
rs1017892533 Chr.16: 790383-790383 on Build GRCh38
rs745347953 Chr.16: 790422-790422 on Build GRCh38
rs751029747 Chr.16: 790408-790408 on Build GRCh38
rs746955988 Chr.16: 790423-790423 on Build GRCh38
rs751930649 Chr.16: 790397-790397 on Build GRCh38
rs767058178 Chr.16: 790316-790316 on Build GRCh38
rs543149772 Chr.16: 790266-790266 on Build GRCh38
rs764092948 Chr.16: 790308-790308 on Build GRCh38
rs768531230 Chr.16: 790377-790377 on Build GRCh38
rs749092374 Chr.16: 790338-790338 on Build GRCh38
rs993753667 Chr.16: 790261-790261 on Build GRCh38
rs372737722 Chr.16: 790245-790245 on Build GRCh38
rs3765263 Chr.16: 790378-790378 on Build GRCh38
rs114140868 Chr.16: 790267-790267 on Build GRCh38
rs776202338 Chr.16: 790392-790392 on Build GRCh38
rs757603349 Chr.16: 790403-790403 on Build GRCh38
rs764526584 Chr.16: 790358-790358 on Build GRCh38
rs536812441 Chr.16: 790418-790418 on Build GRCh38
rs766708697 Chr.16: 790365-790366 on Build GRCh38
rs369085988 Chr.16: 790295-790295 on Build GRCh38
rs755783106 Chr.16: 790317-790317 on Build GRCh38
rs373152305 Chr.16: 790318-790318 on Build GRCh38
rs776146021 Chr.16: 790431-790431 on Build GRCh38
rs753026191 Chr.16: 790319-790319 on Build GRCh38
rs766888701 Chr.16: 790348-790348 on Build GRCh38
rs763648817 Chr.16: 790263-790263 on Build GRCh38
rs746029057 Chr.16: 790329-790329 on Build GRCh38
rs193266504 Chr.16: 790414-790414 on Build GRCh38
rs200891692 Chr.16: 790347-790347 on Build GRCh38
rs778290617 Chr.16: 790288-790288 on Build GRCh38
rs760855561 Chr.16: 790243-790243 on Build GRCh38
rs780364796 Chr.16: 790372-790372 on Build GRCh38
rs764320630 Chr.16: 790396-790396 on Build GRCh38
rs770119809 Chr.16: 790332-790332 on Build GRCh38
rs563011261 Chr.16: 790359-790359 on Build GRCh38
rs902243219 Chr.16: 790282-790282 on Build GRCh38
rs148417318 Chr.16: 790360-790361 on Build GRCh38
rs747567147 Chr.16: 790326-790326 on Build GRCh38
rs775448687 Chr.16: 790303-790303 on Build GRCh38
rs753626456 Chr.16: 790289-790290 on Build GRCh38
rs570353416 Chr.16: 790380-790380 on Build GRCh38
rs368750084 Chr.16: 790258-790258 on Build GRCh38

View Full Product Details