Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 45095610-45095879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUOX2
Entrez Gene ID
50506
Gene Name
dual oxidase 2
Gene Aliases
LNOX2, NOXEF2, P138-TOX, TDH6, THOX2
UniGene
Hs.71377
Gene Chromosome Location
Chr.15: 45092653-45114161 on Build GRCh38
Transcripts
NM_014080.4 XM_005254421.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3501618 c.4048C>T p.P1350S Chr.15: 45095860-45095860 on Build GRCh38
4883019 c.4047C>A p.S1349S Chr.15: 45095861-45095861 on Build GRCh38
244163 c.4055G>A p.G1352D Chr.15: 45095853-45095853 on Build GRCh38
115784 c.4032G>T p.R1344S Chr.15: 45095876-45095876 on Build GRCh38
3887049 c.4046C>T p.S1349F Chr.15: 45095862-45095862 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs554300891 Chr.15: 45095734-45095734 on Build GRCh38
rs371971333 Chr.15: 45095815-45095815 on Build GRCh38
rs1023531092 Chr.15: 45095712-45095712 on Build GRCh38
rs558245016 Chr.15: 45095656-45095656 on Build GRCh38
rs866302157 Chr.15: 45095677-45095677 on Build GRCh38
rs199933166 Chr.15: 45095801-45095801 on Build GRCh38
rs1053545758 Chr.15: 45095697-45095697 on Build GRCh38
rs973189070 Chr.15: 45095809-45095809 on Build GRCh38
rs770824980 Chr.15: 45095631-45095631 on Build GRCh38
rs775001939 Chr.15: 45095853-45095853 on Build GRCh38
rs577507386 Chr.15: 45095759-45095759 on Build GRCh38
rs777812482 Chr.15: 45095827-45095827 on Build GRCh38
rs1003222289 Chr.15: 45095739-45095739 on Build GRCh38
rs750749727 Chr.15: 45095812-45095812 on Build GRCh38
rs16977579 Chr.15: 45095701-45095701 on Build GRCh38
rs370768784 Chr.15: 45095800-45095800 on Build GRCh38
rs775079078 Chr.15: 45095789-45095789 on Build GRCh38
rs760037729 Chr.15: 45095791-45095791 on Build GRCh38
rs374891282 Chr.15: 45095828-45095828 on Build GRCh38
rs576563372 Chr.15: 45095710-45095710 on Build GRCh38
rs774186374 Chr.15: 45095632-45095632 on Build GRCh38
rs745683765 Chr.15: 45095688-45095688 on Build GRCh38
rs774309732 Chr.15: 45095832-45095832 on Build GRCh38
rs777150757 Chr.15: 45095738-45095738 on Build GRCh38
rs568300120 Chr.15: 45095637-45095637 on Build GRCh38
rs913749110 Chr.15: 45095851-45095851 on Build GRCh38
rs754997298 Chr.15: 45095824-45095824 on Build GRCh38
rs534394361 Chr.15: 45095711-45095711 on Build GRCh38
rs115310278 Chr.15: 45095821-45095821 on Build GRCh38
rs944749967 Chr.15: 45095639-45095639 on Build GRCh38
rs771855544 Chr.15: 45095847-45095847 on Build GRCh38
rs747176882 Chr.15: 45095822-45095822 on Build GRCh38
rs903281909 Chr.15: 45095686-45095686 on Build GRCh38
rs745599300 Chr.15: 45095834-45095834 on Build GRCh38
rs367887153 Chr.15: 45095813-45095813 on Build GRCh38
rs759291051 Chr.15: 45095643-45095643 on Build GRCh38
rs767229382 Chr.15: 45095781-45095781 on Build GRCh38
rs977450051 Chr.15: 45095857-45095857 on Build GRCh38
rs1015715892 Chr.15: 45095676-45095676 on Build GRCh38
rs1004375941 Chr.15: 45095657-45095657 on Build GRCh38
rs944676785 Chr.15: 45095852-45095852 on Build GRCh38
rs201798906 Chr.15: 45095810-45095810 on Build GRCh38

View Full Product Details