Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 208160731-208160972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYGA
Entrez Gene ID
1418
Gene Name
crystallin gamma A
Gene Aliases
CRY-g-A, CRYG1, CRYG5
UniGene
Hs.122566
Gene Chromosome Location
Chr.2: 208160740-208163573 on Build GRCh38
Transcripts
NM_014617.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4090671 c.460T>G p.Y154D Chr.2: 208160869-208160869 on Build GRCh38
4949300 c.380T>C p.L127P Chr.2: 208160949-208160949 on Build GRCh38
476828 c.459G>C p.R153S Chr.2: 208160870-208160870 on Build GRCh38
1731790 c.462C>A p.Y154* Chr.2: 208160867-208160867 on Build GRCh38
4674840 c.465C>T p.H155H Chr.2: 208160864-208160864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769374325 Chr.2: 208160885-208160885 on Build GRCh38
rs375520207 Chr.2: 208160761-208160761 on Build GRCh38
rs369021683 Chr.2: 208160947-208160947 on Build GRCh38
rs763049410 Chr.2: 208160886-208160886 on Build GRCh38
rs777116697 Chr.2: 208160756-208160756 on Build GRCh38
rs757259678 Chr.2: 208160881-208160881 on Build GRCh38
rs757258947 Chr.2: 208160753-208160753 on Build GRCh38
rs781323558 Chr.2: 208160883-208160883 on Build GRCh38
rs755519018 Chr.2: 208160936-208160936 on Build GRCh38
rs149711129 Chr.2: 208160816-208160816 on Build GRCh38
rs201789574 Chr.2: 208160766-208160766 on Build GRCh38
rs774234182 Chr.2: 208160934-208160935 on Build GRCh38
rs918346369 Chr.2: 208160750-208160750 on Build GRCh38
rs766462446 Chr.2: 208160863-208160863 on Build GRCh38
rs573473213 Chr.2: 208160910-208160910 on Build GRCh38
rs371947145 Chr.2: 208160928-208160928 on Build GRCh38
rs374969217 Chr.2: 208160937-208160937 on Build GRCh38
rs189542184 Chr.2: 208160778-208160778 on Build GRCh38
rs879113806 Chr.2: 208160837-208160837 on Build GRCh38
rs776270864 Chr.2: 208160813-208160813 on Build GRCh38
rs747567907 Chr.2: 208160811-208160816 on Build GRCh38
rs200186348 Chr.2: 208160823-208160823 on Build GRCh38
rs768077584 Chr.2: 208160854-208160854 on Build GRCh38
rs760632602 Chr.2: 208160909-208160909 on Build GRCh38
rs772644278 Chr.2: 208160852-208160853 on Build GRCh38
rs771924399 Chr.2: 208160776-208160776 on Build GRCh38
rs201293779 Chr.2: 208160870-208160870 on Build GRCh38
rs773791564 Chr.2: 208160906-208160906 on Build GRCh38
rs202138184 Chr.2: 208160884-208160884 on Build GRCh38
rs774692409 Chr.2: 208160899-208160899 on Build GRCh38
rs559628404 Chr.2: 208160904-208160904 on Build GRCh38
rs371873944 Chr.2: 208160836-208160836 on Build GRCh38
rs769312886 Chr.2: 208160819-208160823 on Build GRCh38
rs368405640 Chr.2: 208160815-208160815 on Build GRCh38
rs182000087 Chr.2: 208160940-208160940 on Build GRCh38
rs745819304 Chr.2: 208160930-208160930 on Build GRCh38
rs754121301 Chr.2: 208160864-208160864 on Build GRCh38
rs781260446 Chr.2: 208160754-208160754 on Build GRCh38
rs762293307 Chr.2: 208160853-208160853 on Build GRCh38
rs936862655 Chr.2: 208160817-208160817 on Build GRCh38
rs866669425 Chr.2: 208160839-208160839 on Build GRCh38
rs1025457304 Chr.2: 208160943-208160943 on Build GRCh38
rs200229414 Chr.2: 208160774-208160774 on Build GRCh38
rs756284379 Chr.2: 208160767-208160767 on Build GRCh38
rs746861380 Chr.2: 208160874-208160874 on Build GRCh38
rs764443753 Chr.2: 208160824-208160824 on Build GRCh38
rs757171734 Chr.2: 208160919-208160919 on Build GRCh38
rs542764482 Chr.2: 208160895-208160895 on Build GRCh38
rs201368354 Chr.2: 208160807-208160807 on Build GRCh38
rs746949317 Chr.2: 208160793-208160793 on Build GRCh38
rs140372256 Chr.2: 208160859-208160859 on Build GRCh38
rs980389639 Chr.2: 208160857-208160857 on Build GRCh38
rs780363903 Chr.2: 208160771-208160771 on Build GRCh38
rs986559706 Chr.2: 208160948-208160948 on Build GRCh38
rs759998672 Chr.2: 208160911-208160911 on Build GRCh38
rs762205411 Chr.2: 208160903-208160903 on Build GRCh38
rs777727117 Chr.2: 208160871-208160871 on Build GRCh38
rs369277397 Chr.2: 208160914-208160914 on Build GRCh38
rs1052600801 Chr.2: 208160858-208160858 on Build GRCh38
rs762650691 Chr.2: 208160853-208160853 on Build GRCh38
rs375199678 Chr.2: 208160916-208160916 on Build GRCh38
rs770714825 Chr.2: 208160906-208160906 on Build GRCh38
rs781679000 Chr.2: 208160757-208160758 on Build GRCh38

View Full Product Details