Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 72761793-72762018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIS3
Entrez Gene ID
22894
Gene Name
DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease
Gene Aliases
2810028N01Rik, EXOSC11, KIAA1008, RRP44, dis3p
UniGene
Hs.744104
Gene Chromosome Location
Chr.13: 72755402-72782128 on Build GRCh38
Transcripts
NM_001128226.2 NM_001322348.1 NM_001322349.1 NM_014953.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6971927 c.2335A>T p.I779F Chr.13: 72761930-72761930 on Build GRCh38
6945787 c.2329T>C p.S777P Chr.13: 72761936-72761936 on Build GRCh38
374148 c.2304G>T p.A768A Chr.13: 72761961-72761961 on Build GRCh38
329311 c.2339G>A p.R780K Chr.13: 72761926-72761926 on Build GRCh38
6644260 c.2303C>T p.A768V Chr.13: 72761962-72761962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779298794 Chr.13: 72761972-72761972 on Build GRCh38
rs772600739 Chr.13: 72761842-72761842 on Build GRCh38
rs760792925 Chr.13: 72761844-72761844 on Build GRCh38
rs769002260 Chr.13: 72761823-72761823 on Build GRCh38
rs117383606 Chr.13: 72761962-72761962 on Build GRCh38
rs752456659 Chr.13: 72761899-72761899 on Build GRCh38
rs770256818 Chr.13: 72761880-72761880 on Build GRCh38
rs1015845241 Chr.13: 72761868-72761868 on Build GRCh38
rs920260367 Chr.13: 72761822-72761822 on Build GRCh38
rs748643930 Chr.13: 72761836-72761836 on Build GRCh38
rs764164422 Chr.13: 72761949-72761949 on Build GRCh38
rs969104160 Chr.13: 72761847-72761847 on Build GRCh38
rs766576458 Chr.13: 72761846-72761846 on Build GRCh38
rs751086998 Chr.13: 72761951-72761951 on Build GRCh38
rs756111748 Chr.13: 72761864-72761864 on Build GRCh38
rs937474302 Chr.13: 72761961-72761961 on Build GRCh38
rs756881985 Chr.13: 72761953-72761953 on Build GRCh38
rs773247653 Chr.13: 72761843-72761843 on Build GRCh38
rs370040400 Chr.13: 72761870-72761870 on Build GRCh38
rs780575971 Chr.13: 72761957-72761957 on Build GRCh38
rs191800779 Chr.13: 72761882-72761882 on Build GRCh38
rs758367643 Chr.13: 72761913-72761913 on Build GRCh38
rs774595302 Chr.13: 72761824-72761824 on Build GRCh38
rs776977707 Chr.13: 72761852-72761852 on Build GRCh38
rs759682186 Chr.13: 72761872-72761872 on Build GRCh38

View Full Product Details