Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219281613-219281884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB2
Entrez Gene ID
3300
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
Gene Aliases
CMT2T, DSMA5, HSJ-1, HSJ1, HSPF3
UniGene
Hs.77768
Gene Chromosome Location
Chr.2: 219279318-219286900 on Build GRCh38
Transcripts
NM_001039550.1 NM_006736.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6647106 c.184C>T p.R62W Chr.2: 219281726-219281726 on Build GRCh38
1614392 c.208C>G p.R70G Chr.2: 219281750-219281750 on Build GRCh38
3709424 c.208C>G p.R70G Chr.2: 219281750-219281750 on Build GRCh38
5396271 c.215_216GG>AA p.G72E Chr.2: 219281757-219281758 on Build GRCh38
3577946 c.209G>T p.R70L Chr.2: 219281751-219281751 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755890925 Chr.2: 219281672-219281672 on Build GRCh38
rs3731896 Chr.2: 219281680-219281680 on Build GRCh38
rs922017854 Chr.2: 219281840-219281840 on Build GRCh38
rs749724691 Chr.2: 219281738-219281746 on Build GRCh38
rs1053505349 Chr.2: 219281858-219281858 on Build GRCh38
rs747378005 Chr.2: 219281726-219281726 on Build GRCh38
rs767377598 Chr.2: 219281744-219281744 on Build GRCh38
rs953531733 Chr.2: 219281645-219281645 on Build GRCh38
rs116719245 Chr.2: 219281737-219281737 on Build GRCh38
rs926453312 Chr.2: 219281764-219281764 on Build GRCh38
rs778291801 Chr.2: 219281724-219281724 on Build GRCh38
rs757898570 Chr.2: 219281819-219281819 on Build GRCh38
rs758856704 Chr.2: 219281698-219281698 on Build GRCh38
rs377310796 Chr.2: 219281775-219281775 on Build GRCh38
rs778274050 Chr.2: 219281685-219281686 on Build GRCh38
rs746079387 Chr.2: 219281738-219281738 on Build GRCh38
rs779252318 Chr.2: 219281823-219281824 on Build GRCh38
rs778407121 Chr.2: 219281690-219281690 on Build GRCh38
rs11559157 Chr.2: 219281742-219281742 on Build GRCh38
rs771292719 Chr.2: 219281727-219281727 on Build GRCh38
rs764826178 Chr.2: 219281785-219281785 on Build GRCh38
rs1035188417 Chr.2: 219281707-219281707 on Build GRCh38
rs140172586 Chr.2: 219281741-219281741 on Build GRCh38
rs773179867 Chr.2: 219281750-219281750 on Build GRCh38
rs984882083 Chr.2: 219281673-219281673 on Build GRCh38
rs754628028 Chr.2: 219281780-219281780 on Build GRCh38
rs369512200 Chr.2: 219281693-219281693 on Build GRCh38
rs750218650 Chr.2: 219281671-219281671 on Build GRCh38
rs980692062 Chr.2: 219281761-219281761 on Build GRCh38
rs752224503 Chr.2: 219281815-219281815 on Build GRCh38
rs111812148 Chr.2: 219281717-219281717 on Build GRCh38
rs756458971 Chr.2: 219281682-219281685 on Build GRCh38
rs766643040 Chr.2: 219281805-219281819 on Build GRCh38
rs1005659124 Chr.2: 219281653-219281653 on Build GRCh38
rs730882139 Chr.2: 219281772-219281772 on Build GRCh38
rs753570551 Chr.2: 219281773-219281773 on Build GRCh38
rs1003266824 Chr.2: 219281703-219281703 on Build GRCh38
rs771352242 Chr.2: 219281811-219281811 on Build GRCh38
rs949341803 Chr.2: 219281745-219281745 on Build GRCh38
rs766183158 Chr.2: 219281751-219281751 on Build GRCh38
rs554986026 Chr.2: 219281852-219281852 on Build GRCh38
rs765997763 Chr.2: 219281677-219281677 on Build GRCh38

View Full Product Details