Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144096985-144097256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYC1
Entrez Gene ID
1537
Gene Name
cytochrome c1
Gene Aliases
MC3DN6, UQCR4
UniGene
Hs.289271
Gene Chromosome Location
Chr.8: 144095035-144097527 on Build GRCh38
Transcripts
NM_001916.4 XM_017013102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6027591 c.860G>A p.R287H Chr.8: 144097121-144097121 on Build GRCh38
1755627 c.853C>T p.R285* Chr.8: 144097114-144097114 on Build GRCh38
5436527 c.870C>G p.L290L Chr.8: 144097131-144097131 on Build GRCh38
749635 c.795G>C p.Q265H Chr.8: 144097056-144097056 on Build GRCh38
7079609 c.804G>C p.K268N Chr.8: 144097065-144097065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369289950 Chr.8: 144097154-144097154 on Build GRCh38
rs764152487 Chr.8: 144097215-144097215 on Build GRCh38
rs753008933 Chr.8: 144097236-144097238 on Build GRCh38
rs375101616 Chr.8: 144097095-144097095 on Build GRCh38
rs374190143 Chr.8: 144097194-144097194 on Build GRCh38
rs371923821 Chr.8: 144097068-144097068 on Build GRCh38
rs547540376 Chr.8: 144097069-144097069 on Build GRCh38
rs765197336 Chr.8: 144097233-144097235 on Build GRCh38
rs755658614 Chr.8: 144097160-144097160 on Build GRCh38
rs769234556 Chr.8: 144097123-144097123 on Build GRCh38
rs771105454 Chr.8: 144097199-144097199 on Build GRCh38
rs1056904550 Chr.8: 144097114-144097114 on Build GRCh38
rs761709043 Chr.8: 144097229-144097229 on Build GRCh38
rs769287590 Chr.8: 144097177-144097177 on Build GRCh38
rs756988987 Chr.8: 144097152-144097152 on Build GRCh38
rs776699655 Chr.8: 144097200-144097200 on Build GRCh38
rs34755830 Chr.8: 144097221-144097221 on Build GRCh38
rs745484298 Chr.8: 144097161-144097161 on Build GRCh38
rs774809212 Chr.8: 144097125-144097125 on Build GRCh38
rs576800870 Chr.8: 144097190-144097190 on Build GRCh38
rs199860966 Chr.8: 144097189-144097189 on Build GRCh38
rs1126919 Chr.8: 144097094-144097094 on Build GRCh38
rs746744231 Chr.8: 144097083-144097083 on Build GRCh38
rs74494160 Chr.8: 144097120-144097120 on Build GRCh38
rs776238504 Chr.8: 144097006-144097006 on Build GRCh38
rs200281879 Chr.8: 144097141-144097141 on Build GRCh38
rs532256208 Chr.8: 144097016-144097016 on Build GRCh38
rs372859144 Chr.8: 144097197-144097197 on Build GRCh38
rs535993678 Chr.8: 144097128-144097128 on Build GRCh38
rs758242318 Chr.8: 144097070-144097070 on Build GRCh38
rs780653499 Chr.8: 144097085-144097085 on Build GRCh38
rs569607010 Chr.8: 144097143-144097143 on Build GRCh38
rs745441729 Chr.8: 144097108-144097108 on Build GRCh38
rs377674285 Chr.8: 144097217-144097217 on Build GRCh38
rs776707883 Chr.8: 144097208-144097209 on Build GRCh38
rs761771812 Chr.8: 144097187-144097187 on Build GRCh38
rs768546404 Chr.8: 144097010-144097010 on Build GRCh38
rs767193571 Chr.8: 144097232-144097232 on Build GRCh38
rs777570129 Chr.8: 144097072-144097072 on Build GRCh38
rs775464970 Chr.8: 144097035-144097035 on Build GRCh38
rs138756019 Chr.8: 144097050-144097050 on Build GRCh38
rs755733810 Chr.8: 144097168-144097168 on Build GRCh38
rs376851349 Chr.8: 144097182-144097182 on Build GRCh38
rs762013973 Chr.8: 144097126-144097126 on Build GRCh38
rs367892680 Chr.8: 144097048-144097048 on Build GRCh38
rs758366796 Chr.8: 144097145-144097145 on Build GRCh38
rs973940506 Chr.8: 144097046-144097046 on Build GRCh38
rs749790299 Chr.8: 144097121-144097121 on Build GRCh38
rs769424627 Chr.8: 144097115-144097115 on Build GRCh38
rs774085819 Chr.8: 144097149-144097150 on Build GRCh38
rs112492233 Chr.8: 144097032-144097032 on Build GRCh38
rs765213313 Chr.8: 144097059-144097059 on Build GRCh38
rs763477511 Chr.8: 144097019-144097019 on Build GRCh38
rs748766177 Chr.8: 144097175-144097175 on Build GRCh38
rs374936583 Chr.8: 144097025-144097025 on Build GRCh38
rs1006594181 Chr.8: 144097206-144097206 on Build GRCh38
rs375851599 Chr.8: 144097151-144097151 on Build GRCh38
rs760788386 Chr.8: 144097137-144097137 on Build GRCh38
rs756125106 Chr.8: 144097053-144097053 on Build GRCh38
rs942669652 Chr.8: 144097183-144097183 on Build GRCh38
rs767379790 Chr.8: 144097204-144097205 on Build GRCh38
rs759722273 Chr.8: 144097228-144097228 on Build GRCh38
rs372487965 Chr.8: 144097169-144097169 on Build GRCh38
rs770438704 Chr.8: 144097010-144097010 on Build GRCh38
rs369102230 Chr.8: 144097186-144097186 on Build GRCh38
rs370195911 Chr.8: 144097024-144097024 on Build GRCh38
rs2070688 Chr.8: 144097007-144097007 on Build GRCh38
rs370734008 Chr.8: 144097235-144097235 on Build GRCh38
rs368304577 Chr.8: 144097107-144097107 on Build GRCh38
rs756940609 Chr.8: 144097084-144097084 on Build GRCh38
rs763872484 Chr.8: 144097146-144097146 on Build GRCh38
rs759556070 Chr.8: 144097205-144097205 on Build GRCh38
rs773290081 Chr.8: 144097131-144097131 on Build GRCh38
rs1043903915 Chr.8: 144097165-144097165 on Build GRCh38
rs777401702 Chr.8: 144097173-144097173 on Build GRCh38

View Full Product Details