Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 12815278-12815505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAND2
Entrez Gene ID
23066
Gene Name
cullin associated and neddylation dissociated 2 (putative)
Gene Aliases
TIP120B, Tp120b
UniGene
Hs.343664
Gene Chromosome Location
Chr.3: 12796654-12834818 on Build GRCh38
Transcripts
NM_001162499.1 NM_012298.2 XM_011533503.2 XM_011533504.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3392023 c.1172A>T p.N391I Chr.3: 12815306-12815306 on Build GRCh38
3408207 c.958T>A p.W320R Chr.3: 12815371-12815371 on Build GRCh38
6317916 c.1020+7G>T p.? Chr.3: 12815440-12815440 on Build GRCh38
1136928 c.1251G>T p.M417I Chr.3: 12815385-12815385 on Build GRCh38
3408206 c.1237T>A p.W413R Chr.3: 12815371-12815371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769635010 Chr.3: 12815429-12815429 on Build GRCh38
rs752676710 Chr.3: 12815338-12815338 on Build GRCh38
rs531827651 Chr.3: 12815427-12815427 on Build GRCh38
rs367907914 Chr.3: 12815319-12815319 on Build GRCh38
rs372471540 Chr.3: 12815355-12815355 on Build GRCh38
rs760934303 Chr.3: 12815368-12815368 on Build GRCh38
rs201177236 Chr.3: 12815358-12815358 on Build GRCh38
rs1055702063 Chr.3: 12815326-12815326 on Build GRCh38
rs757999829 Chr.3: 12815337-12815337 on Build GRCh38
rs762134748 Chr.3: 12815453-12815453 on Build GRCh38
rs761175364 Chr.3: 12815442-12815442 on Build GRCh38
rs993019070 Chr.3: 12815425-12815425 on Build GRCh38
rs756041832 Chr.3: 12815466-12815466 on Build GRCh38
rs530110068 Chr.3: 12815403-12815403 on Build GRCh38
rs767082153 Chr.3: 12815464-12815464 on Build GRCh38
rs549950370 Chr.3: 12815457-12815457 on Build GRCh38
rs777006257 Chr.3: 12815449-12815449 on Build GRCh38
rs147467944 Chr.3: 12815389-12815389 on Build GRCh38
rs769701239 Chr.3: 12815356-12815356 on Build GRCh38
rs779090145 Chr.3: 12815475-12815475 on Build GRCh38
rs750146696 Chr.3: 12815335-12815335 on Build GRCh38
rs766098123 Chr.3: 12815454-12815454 on Build GRCh38
rs547890865 Chr.3: 12815302-12815302 on Build GRCh38
rs764459329 Chr.3: 12815373-12815373 on Build GRCh38
rs62637645 Chr.3: 12815404-12815404 on Build GRCh38
rs754156396 Chr.3: 12815383-12815383 on Build GRCh38
rs374837362 Chr.3: 12815307-12815307 on Build GRCh38
rs544160485 Chr.3: 12815330-12815330 on Build GRCh38
rs767977141 Chr.3: 12815363-12815363 on Build GRCh38
rs764539196 Chr.3: 12815448-12815448 on Build GRCh38
rs575226919 Chr.3: 12815315-12815315 on Build GRCh38
rs746550726 Chr.3: 12815347-12815347 on Build GRCh38
rs771117872 Chr.3: 12815438-12815438 on Build GRCh38
rs2305398 Chr.3: 12815357-12815357 on Build GRCh38
rs890250051 Chr.3: 12815334-12815334 on Build GRCh38
rs777561017 Chr.3: 12815434-12815434 on Build GRCh38
rs758031161 Chr.3: 12815392-12815392 on Build GRCh38
rs774361974 Chr.3: 12815360-12815360 on Build GRCh38
rs749076921 Chr.3: 12815471-12815471 on Build GRCh38
rs372664387 Chr.3: 12815440-12815440 on Build GRCh38
rs1041656149 Chr.3: 12815400-12815400 on Build GRCh38
rs754576042 Chr.3: 12815413-12815413 on Build GRCh38
rs781329525 Chr.3: 12815348-12815348 on Build GRCh38
rs752156092 Chr.3: 12815465-12815465 on Build GRCh38
rs751324030 Chr.3: 12815455-12815455 on Build GRCh38
rs753034201 Chr.3: 12815322-12815322 on Build GRCh38
rs759658409 Chr.3: 12815361-12815361 on Build GRCh38
rs746031937 Chr.3: 12815481-12815481 on Build GRCh38
rs781275010 Chr.3: 12815417-12815417 on Build GRCh38
rs140978687 Chr.3: 12815439-12815439 on Build GRCh38
rs777617837 Chr.3: 12815470-12815470 on Build GRCh38
rs539271019 Chr.3: 12815298-12815298 on Build GRCh38
rs749056146 Chr.3: 12815435-12815435 on Build GRCh38
rs754526582 Chr.3: 12815460-12815460 on Build GRCh38
rs564884630 Chr.3: 12815299-12815299 on Build GRCh38
rs765355002 Chr.3: 12815296-12815296 on Build GRCh38
rs756537260 Chr.3: 12815323-12815323 on Build GRCh38
rs757106017 Chr.3: 12815474-12815474 on Build GRCh38
rs552901240 Chr.3: 12815459-12815459 on Build GRCh38
rs748178340 Chr.3: 12815420-12815420 on Build GRCh38
rs751654103 Chr.3: 12815308-12815308 on Build GRCh38
rs780373526 Chr.3: 12815353-12815354 on Build GRCh38

View Full Product Details