Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 80159685-80159950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAH
Entrez Gene ID
2184
Gene Name
fumarylacetoacetate hydrolase
Gene Aliases
-
UniGene
Hs.73875
Gene Chromosome Location
Chr.15: 80152891-80186582 on Build GRCh38
Transcripts
NM_000137.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7424486 c.224A>T p.Q75L Chr.15: 80159787-80159787 on Build GRCh38
2011604 c.231C>G p.A77A Chr.15: 80159794-80159794 on Build GRCh38
6657979 c.237G>A p.K79K Chr.15: 80159800-80159800 on Build GRCh38
965502 c.197C>T p.T66I Chr.15: 80159760-80159760 on Build GRCh38
2011607 c.301G>A p.E101K Chr.15: 80159864-80159864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547847694 Chr.15: 80159859-80159859 on Build GRCh38
rs371012820 Chr.15: 80159891-80159891 on Build GRCh38
rs758347578 Chr.15: 80159865-80159865 on Build GRCh38
rs36122289 Chr.15: 80159806-80159806 on Build GRCh38
rs1057516408 Chr.15: 80159878-80159878 on Build GRCh38
rs775248570 Chr.15: 80159818-80159818 on Build GRCh38
rs545954407 Chr.15: 80159774-80159774 on Build GRCh38
rs956885219 Chr.15: 80159729-80159729 on Build GRCh38
rs750111883 Chr.15: 80159725-80159725 on Build GRCh38
rs754226189 Chr.15: 80159854-80159854 on Build GRCh38
rs1021168670 Chr.15: 80159737-80159737 on Build GRCh38
rs751008263 Chr.15: 80159712-80159712 on Build GRCh38
rs748140237 Chr.15: 80159776-80159776 on Build GRCh38
rs763985544 Chr.15: 80159824-80159824 on Build GRCh38
rs772218801 Chr.15: 80159787-80159787 on Build GRCh38
rs975262013 Chr.15: 80159855-80159855 on Build GRCh38
rs749729119 Chr.15: 80159739-80159739 on Build GRCh38
rs11555097 Chr.15: 80159864-80159864 on Build GRCh38
rs562770334 Chr.15: 80159730-80159730 on Build GRCh38
rs929155893 Chr.15: 80159895-80159895 on Build GRCh38
rs756489499 Chr.15: 80159727-80159727 on Build GRCh38
rs771205853 Chr.15: 80159814-80159814 on Build GRCh38
rs747145977 Chr.15: 80159871-80159871 on Build GRCh38
rs749045075 Chr.15: 80159888-80159888 on Build GRCh38
rs777899653 Chr.15: 80159870-80159870 on Build GRCh38
rs374371839 Chr.15: 80159834-80159834 on Build GRCh38
rs746381874 Chr.15: 80159731-80159731 on Build GRCh38
rs779516322 Chr.15: 80159764-80159764 on Build GRCh38
rs766184800 Chr.15: 80159853-80159853 on Build GRCh38
rs767412192 Chr.15: 80159903-80159903 on Build GRCh38
rs755815616 Chr.15: 80159847-80159847 on Build GRCh38
rs772755729 Chr.15: 80159912-80159912 on Build GRCh38
rs1370274 Chr.15: 80159733-80159733 on Build GRCh38
rs771117713 Chr.15: 80159872-80159872 on Build GRCh38
rs769265153 Chr.15: 80159742-80159742 on Build GRCh38
rs192467738 Chr.15: 80159724-80159724 on Build GRCh38
rs768520008 Chr.15: 80159897-80159897 on Build GRCh38
rs370095143 Chr.15: 80159812-80159812 on Build GRCh38
rs748750943 Chr.15: 80159881-80159881 on Build GRCh38
rs965080831 Chr.15: 80159845-80159845 on Build GRCh38
rs567587933 Chr.15: 80159863-80159863 on Build GRCh38
rs776875979 Chr.15: 80159882-80159882 on Build GRCh38
rs33929922 Chr.15: 80159830-80159830 on Build GRCh38
rs1370273 Chr.15: 80159926-80159926 on Build GRCh38
rs374617446 Chr.15: 80159899-80159899 on Build GRCh38
rs749967814 Chr.15: 80159840-80159840 on Build GRCh38
rs762783017 Chr.15: 80159819-80159819 on Build GRCh38
rs772531438 Chr.15: 80159897-80159897 on Build GRCh38
rs764073453 Chr.15: 80159711-80159711 on Build GRCh38
rs200373686 Chr.15: 80159720-80159720 on Build GRCh38
rs952662827 Chr.15: 80159886-80159886 on Build GRCh38
rs897644297 Chr.15: 80159889-80159889 on Build GRCh38

View Full Product Details