Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58528254-58528527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT1
Entrez Gene ID
23019
Gene Name
CCR4-NOT transcription complex subunit 1
Gene Aliases
AD-005, CDC39, NOT1, NOT1H
UniGene
Hs.460923
Gene Chromosome Location
Chr.16: 58519946-58629886 on Build GRCh38
Transcripts
NM_001265612.1 NM_016284.4 NR_049763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5851136 c.6433C>T p.P2145S Chr.16: 58528495-58528495 on Build GRCh38
7330900 c.6426C>T p.L2142L Chr.16: 58528502-58528502 on Build GRCh38
4503478 c.6405C>T p.A2135A Chr.16: 58528523-58528523 on Build GRCh38
2156321 c.6447delT p.L2150fs*1 Chr.16: 58528481-58528481 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778089070 Chr.16: 58528449-58528449 on Build GRCh38
rs1000900366 Chr.16: 58528418-58528418 on Build GRCh38
rs765902789 Chr.16: 58528342-58528342 on Build GRCh38
rs775821602 Chr.16: 58528471-58528471 on Build GRCh38
rs928996841 Chr.16: 58528327-58528327 on Build GRCh38
rs1014163838 Chr.16: 58528407-58528407 on Build GRCh38
rs74325935 Chr.16: 58528385-58528385 on Build GRCh38
rs772446701 Chr.16: 58528303-58528303 on Build GRCh38
rs771154365 Chr.16: 58528324-58528324 on Build GRCh38
rs753412609 Chr.16: 58528430-58528431 on Build GRCh38
rs529026281 Chr.16: 58528411-58528411 on Build GRCh38
rs781413678 Chr.16: 58528459-58528459 on Build GRCh38
rs750406221 Chr.16: 58528377-58528377 on Build GRCh38
rs767880246 Chr.16: 58528301-58528302 on Build GRCh38
rs755811720 Chr.16: 58528338-58528340 on Build GRCh38
rs770207635 Chr.16: 58528354-58528354 on Build GRCh38
rs919171853 Chr.16: 58528317-58528317 on Build GRCh38
rs373218331 Chr.16: 58528472-58528472 on Build GRCh38
rs767326137 Chr.16: 58528376-58528376 on Build GRCh38
rs753423118 Chr.16: 58528395-58528395 on Build GRCh38
rs377330465 Chr.16: 58528499-58528499 on Build GRCh38
rs778597416 Chr.16: 58528442-58528442 on Build GRCh38
rs576609053 Chr.16: 58528417-58528417 on Build GRCh38
rs778396152 Chr.16: 58528308-58528308 on Build GRCh38
rs189458094 Chr.16: 58528463-58528463 on Build GRCh38
rs199925728 Chr.16: 58528282-58528282 on Build GRCh38
rs543756827 Chr.16: 58528454-58528454 on Build GRCh38
rs759820619 Chr.16: 58528295-58528296 on Build GRCh38
rs763703416 Chr.16: 58528362-58528365 on Build GRCh38
rs202006884 Chr.16: 58528505-58528505 on Build GRCh38
rs867743974 Chr.16: 58528440-58528440 on Build GRCh38
rs748617437 Chr.16: 58528301-58528301 on Build GRCh38
rs1034485147 Chr.16: 58528402-58528402 on Build GRCh38
rs776931988 Chr.16: 58528336-58528336 on Build GRCh38
rs142254978 Chr.16: 58528375-58528375 on Build GRCh38
rs754623907 Chr.16: 58528430-58528430 on Build GRCh38
rs896619477 Chr.16: 58528406-58528406 on Build GRCh38
rs774001587 Chr.16: 58528370-58528370 on Build GRCh38
rs902763301 Chr.16: 58528415-58528415 on Build GRCh38
rs374768844 Chr.16: 58528443-58528443 on Build GRCh38
rs758841492 Chr.16: 58528444-58528444 on Build GRCh38
rs747017938 Chr.16: 58528323-58528323 on Build GRCh38
rs746001672 Chr.16: 58528460-58528460 on Build GRCh38
rs74530059 Chr.16: 58528456-58528456 on Build GRCh38
rs752365892 Chr.16: 58528303-58528303 on Build GRCh38

View Full Product Details