Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81252750-81252989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKMT2
Entrez Gene ID
1160
Gene Name
creatine kinase, mitochondrial 2
Gene Aliases
SMTCK
UniGene
Hs.80691
Gene Chromosome Location
Chr.5: 81233320-81266398 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

Gene Symbol
CKMT2-AS1
Entrez Gene ID
100131067
Gene Name
CKMT2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655855
Gene Chromosome Location
Chr.5: 81237565-81301569 on Build GRCh38
Transcripts
NM_001099735.1 NM_001099736.1 NM_001825.2 NR_034121.1 NR_034122.1 NR_034123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7102734 c.273C>G p.I91M Chr.5: 81252815-81252815 on Build GRCh38
7177377 c.248A>C p.N83T Chr.5: 81252790-81252790 on Build GRCh38
739056 c.213C>A p.A71A Chr.5: 81252755-81252755 on Build GRCh38
294248 c.211G>A p.A71T Chr.5: 81252753-81252753 on Build GRCh38
3856642 c.319G>T p.G107C Chr.5: 81252861-81252861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969137305 Chr.5: 81252806-81252806 on Build GRCh38
rs539699710 Chr.5: 81252891-81252891 on Build GRCh38
rs750520528 Chr.5: 81252857-81252857 on Build GRCh38
rs112184173 Chr.5: 81252824-81252824 on Build GRCh38
rs201684795 Chr.5: 81252938-81252938 on Build GRCh38
rs746721484 Chr.5: 81252792-81252792 on Build GRCh38
rs772924692 Chr.5: 81252791-81252791 on Build GRCh38
rs148799671 Chr.5: 81252856-81252856 on Build GRCh38
rs550930859 Chr.5: 81252843-81252843 on Build GRCh38
rs759062118 Chr.5: 81252808-81252808 on Build GRCh38
rs552196484 Chr.5: 81252800-81252800 on Build GRCh38
rs770539483 Chr.5: 81252787-81252787 on Build GRCh38
rs998092775 Chr.5: 81252775-81252775 on Build GRCh38
rs751963609 Chr.5: 81252842-81252842 on Build GRCh38
rs372080856 Chr.5: 81252913-81252913 on Build GRCh38
rs570195719 Chr.5: 81252949-81252949 on Build GRCh38
rs756242601 Chr.5: 81252859-81252859 on Build GRCh38
rs770566266 Chr.5: 81252799-81252799 on Build GRCh38
rs1021926867 Chr.5: 81252790-81252790 on Build GRCh38
rs367843661 Chr.5: 81252879-81252879 on Build GRCh38
rs772843767 Chr.5: 81252857-81252858 on Build GRCh38
rs746774404 Chr.5: 81252908-81252908 on Build GRCh38
rs199560112 Chr.5: 81252889-81252889 on Build GRCh38
rs111899125 Chr.5: 81252909-81252909 on Build GRCh38
rs778884873 Chr.5: 81252772-81252772 on Build GRCh38
rs140687986 Chr.5: 81252771-81252771 on Build GRCh38
rs770617324 Chr.5: 81252928-81252928 on Build GRCh38
rs764600725 Chr.5: 81252815-81252815 on Build GRCh38
rs779958865 Chr.5: 81252878-81252878 on Build GRCh38
rs993071244 Chr.5: 81252886-81252886 on Build GRCh38
rs777611301 Chr.5: 81252895-81252895 on Build GRCh38
rs367749573 Chr.5: 81252778-81252778 on Build GRCh38
rs764562887 Chr.5: 81252834-81252834 on Build GRCh38
rs144313442 Chr.5: 81252785-81252785 on Build GRCh38

View Full Product Details