Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2443474-2443715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNF
Entrez Gene ID
899
Gene Name
cyclin F
Gene Aliases
FBX1, FBXO1
UniGene
Hs.1973
Gene Chromosome Location
Chr.16: 2425438-2458858 on Build GRCh38
Transcripts
NM_001323538.1 NM_001761.2 XM_017023822.1 XM_017023823.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1478698 c.843G>C p.E281D Chr.16: 2443714-2443714 on Build GRCh38
4748669 c.821T>A p.L274Q Chr.16: 2443692-2443692 on Build GRCh38
6478646 c.823G>C p.E275Q Chr.16: 2443694-2443694 on Build GRCh38
6538426 c.812A>G p.Q271R Chr.16: 2443683-2443683 on Build GRCh38
4591576 c.839G>A p.S280N Chr.16: 2443710-2443710 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762716472 Chr.16: 2443513-2443513 on Build GRCh38
rs145432131 Chr.16: 2443622-2443622 on Build GRCh38
rs374485469 Chr.16: 2443602-2443602 on Build GRCh38
rs770551533 Chr.16: 2443619-2443619 on Build GRCh38
rs764227871 Chr.16: 2443681-2443681 on Build GRCh38
rs747519392 Chr.16: 2443636-2443637 on Build GRCh38
rs200870073 Chr.16: 2443623-2443623 on Build GRCh38
rs769286504 Chr.16: 2443608-2443608 on Build GRCh38
rs775441275 Chr.16: 2443627-2443627 on Build GRCh38
rs192314535 Chr.16: 2443668-2443668 on Build GRCh38
rs376294554 Chr.16: 2443618-2443618 on Build GRCh38
rs760553008 Chr.16: 2443636-2443636 on Build GRCh38
rs946673080 Chr.16: 2443537-2443537 on Build GRCh38
rs940613269 Chr.16: 2443584-2443584 on Build GRCh38
rs772064770 Chr.16: 2443601-2443602 on Build GRCh38
rs899872820 Chr.16: 2443569-2443569 on Build GRCh38
rs780647051 Chr.16: 2443603-2443604 on Build GRCh38
rs762030951 Chr.16: 2443690-2443690 on Build GRCh38
rs371078328 Chr.16: 2443600-2443600 on Build GRCh38
rs769349067 Chr.16: 2443638-2443640 on Build GRCh38
rs998250590 Chr.16: 2443571-2443571 on Build GRCh38
rs1039699003 Chr.16: 2443544-2443544 on Build GRCh38
rs537427531 Chr.16: 2443598-2443598 on Build GRCh38
rs753937291 Chr.16: 2443683-2443683 on Build GRCh38
rs909160973 Chr.16: 2443574-2443574 on Build GRCh38
rs768511877 Chr.16: 2443514-2443514 on Build GRCh38
rs867407362 Chr.16: 2443646-2443646 on Build GRCh38
rs747471774 Chr.16: 2443607-2443607 on Build GRCh38
rs774051087 Chr.16: 2443620-2443620 on Build GRCh38

View Full Product Details