Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 55114091-55114364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC20B
Entrez Gene ID
166979
Gene Name
cell division cycle 20B
Gene Aliases
G6VTS76519
UniGene
Hs.669184
Gene Chromosome Location
Chr.5: 55112971-55173177 on Build GRCh38
Transcripts
NM_001145734.2 NM_001170402.1 NM_152623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6898325 c.1530T>A p.S510S Chr.5: 55114236-55114236 on Build GRCh38
149836 c.1495C>T p.R499W Chr.5: 55114271-55114271 on Build GRCh38
3761260 c.1381C>T p.R461W Chr.5: 55114271-55114271 on Build GRCh38
4812343 c.1383G>A p.R461R Chr.5: 55114269-55114269 on Build GRCh38
1311209 c.1497G>A p.R499R Chr.5: 55114269-55114269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771475794 Chr.5: 55114262-55114262 on Build GRCh38
rs779710949 Chr.5: 55114320-55114321 on Build GRCh38
rs3104230 Chr.5: 55114292-55114292 on Build GRCh38
rs996265912 Chr.5: 55114145-55114145 on Build GRCh38
rs565721621 Chr.5: 55114147-55114147 on Build GRCh38
rs1010248575 Chr.5: 55114255-55114255 on Build GRCh38
rs777807464 Chr.5: 55114324-55114324 on Build GRCh38
rs148916672 Chr.5: 55114222-55114222 on Build GRCh38
rs375314617 Chr.5: 55114221-55114221 on Build GRCh38
rs908137977 Chr.5: 55114119-55114119 on Build GRCh38
rs748408984 Chr.5: 55114249-55114249 on Build GRCh38
rs76668286 Chr.5: 55114125-55114125 on Build GRCh38
rs750368816 Chr.5: 55114235-55114235 on Build GRCh38
rs778966002 Chr.5: 55114245-55114245 on Build GRCh38
rs372402086 Chr.5: 55114243-55114243 on Build GRCh38
rs747510518 Chr.5: 55114261-55114261 on Build GRCh38
rs758714878 Chr.5: 55114321-55114321 on Build GRCh38
rs146563953 Chr.5: 55114209-55114209 on Build GRCh38
rs369841777 Chr.5: 55114224-55114224 on Build GRCh38
rs866941018 Chr.5: 55114156-55114156 on Build GRCh38
rs1040784614 Chr.5: 55114306-55114306 on Build GRCh38
rs952296180 Chr.5: 55114157-55114157 on Build GRCh38
rs761184916 Chr.5: 55114179-55114179 on Build GRCh38
rs444527 Chr.5: 55114271-55114271 on Build GRCh38
rs753861241 Chr.5: 55114309-55114309 on Build GRCh38
rs778784440 Chr.5: 55114319-55114319 on Build GRCh38
rs748305718 Chr.5: 55114320-55114320 on Build GRCh38
rs746018437 Chr.5: 55114241-55114241 on Build GRCh38
rs773338405 Chr.5: 55114254-55114254 on Build GRCh38
rs961095671 Chr.5: 55114214-55114214 on Build GRCh38
rs762644545 Chr.5: 55114218-55114218 on Build GRCh38
rs76966008 Chr.5: 55114127-55114127 on Build GRCh38
rs749147711 Chr.5: 55114242-55114242 on Build GRCh38
rs779967998 Chr.5: 55114239-55114239 on Build GRCh38
rs767127067 Chr.5: 55114277-55114277 on Build GRCh38
rs771639463 Chr.5: 55114312-55114313 on Build GRCh38
rs754663488 Chr.5: 55114311-55114311 on Build GRCh38
rs777145572 Chr.5: 55114270-55114270 on Build GRCh38
rs371195518 Chr.5: 55114302-55114302 on Build GRCh38
rs372227357 Chr.5: 55114207-55114207 on Build GRCh38
rs369106085 Chr.5: 55114233-55114233 on Build GRCh38
rs141310624 Chr.5: 55114237-55114237 on Build GRCh38
rs773815311 Chr.5: 55114272-55114272 on Build GRCh38
rs150345313 Chr.5: 55114325-55114325 on Build GRCh38
rs200898156 Chr.5: 55114253-55114253 on Build GRCh38
rs756103007 Chr.5: 55114281-55114281 on Build GRCh38

View Full Product Details