Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158770703-158770958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EZR
Entrez Gene ID
7430
Gene Name
ezrin
Gene Aliases
CVIL, CVL, HEL-S-105, VIL2
UniGene
Hs.487027
Gene Chromosome Location
Chr.6: 158765741-158819424 on Build GRCh38
Transcripts
NM_001111077.1 NM_003379.4 XM_011536110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3396634 c.1049G>A p.R350Q Chr.6: 158770805-158770805 on Build GRCh38
5497381 c.1070A>C p.K357T Chr.6: 158770784-158770784 on Build GRCh38
4003772 c.1090+8G>A p.? Chr.6: 158770756-158770756 on Build GRCh38
1251650 c.996C>T p.T332T Chr.6: 158770858-158770858 on Build GRCh38
7157300 c.1057G>A p.D353N Chr.6: 158770797-158770797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779824806 Chr.6: 158770913-158770913 on Build GRCh38
rs3212306 Chr.6: 158770929-158770929 on Build GRCh38
rs183636404 Chr.6: 158770858-158770858 on Build GRCh38
rs1016073887 Chr.6: 158770868-158770868 on Build GRCh38
rs768425876 Chr.6: 158770831-158770833 on Build GRCh38
rs375840157 Chr.6: 158770857-158770857 on Build GRCh38
rs762239919 Chr.6: 158770844-158770845 on Build GRCh38
rs112036858 Chr.6: 158770897-158770897 on Build GRCh38
rs751963489 Chr.6: 158770894-158770894 on Build GRCh38
rs779446669 Chr.6: 158770783-158770783 on Build GRCh38
rs530513639 Chr.6: 158770757-158770757 on Build GRCh38
rs35651592 Chr.6: 158770732-158770733 on Build GRCh38
rs771890889 Chr.6: 158770936-158770936 on Build GRCh38
rs3212308 Chr.6: 158770756-158770756 on Build GRCh38
rs78196406 Chr.6: 158770792-158770792 on Build GRCh38
rs755883473 Chr.6: 158770901-158770901 on Build GRCh38
rs751186941 Chr.6: 158770804-158770804 on Build GRCh38
rs748493999 Chr.6: 158770786-158770786 on Build GRCh38
rs774292855 Chr.6: 158770819-158770819 on Build GRCh38
rs202086677 Chr.6: 158770849-158770849 on Build GRCh38
rs748863915 Chr.6: 158770822-158770824 on Build GRCh38
rs750597189 Chr.6: 158770732-158770732 on Build GRCh38
rs748180011 Chr.6: 158770935-158770935 on Build GRCh38
rs774517856 Chr.6: 158770843-158770844 on Build GRCh38
rs772879132 Chr.6: 158770828-158770828 on Build GRCh38
rs3212307 Chr.6: 158770933-158770933 on Build GRCh38
rs1056973 Chr.6: 158770845-158770845 on Build GRCh38
rs780721379 Chr.6: 158770793-158770793 on Build GRCh38
rs745450636 Chr.6: 158770796-158770796 on Build GRCh38
rs370931420 Chr.6: 158770741-158770741 on Build GRCh38
rs886646450 Chr.6: 158770934-158770934 on Build GRCh38
rs769570013 Chr.6: 158770800-158770800 on Build GRCh38
rs767272547 Chr.6: 158770823-158770823 on Build GRCh38
rs778716661 Chr.6: 158770921-158770921 on Build GRCh38
rs567350960 Chr.6: 158770807-158770807 on Build GRCh38
rs112705375 Chr.6: 158770896-158770896 on Build GRCh38
rs753956634 Chr.6: 158770841-158770841 on Build GRCh38
rs769455167 Chr.6: 158770747-158770748 on Build GRCh38
rs748827570 Chr.6: 158770917-158770917 on Build GRCh38
rs370369131 Chr.6: 158770824-158770824 on Build GRCh38
rs1056974 Chr.6: 158770844-158770844 on Build GRCh38
rs552845390 Chr.6: 158770731-158770731 on Build GRCh38
rs761523413 Chr.6: 158770820-158770820 on Build GRCh38
rs775200943 Chr.6: 158770805-158770805 on Build GRCh38
rs755662102 Chr.6: 158770830-158770830 on Build GRCh38
rs141741749 Chr.6: 158770806-158770806 on Build GRCh38
rs765685742 Chr.6: 158770855-158770855 on Build GRCh38
rs374945393 Chr.6: 158770829-158770829 on Build GRCh38
rs778271542 Chr.6: 158770865-158770865 on Build GRCh38
rs756186576 Chr.6: 158770735-158770735 on Build GRCh38
rs775192844 Chr.6: 158770816-158770818 on Build GRCh38
rs758312834 Chr.6: 158770875-158770876 on Build GRCh38

View Full Product Details