Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186275732-186275967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F11
Entrez Gene ID
2160
Gene Name
coagulation factor XI
Gene Aliases
FXI
UniGene
Hs.1430
Gene Chromosome Location
Chr.4: 186265945-186290727 on Build GRCh38
Transcripts
NM_000128.3 XM_005262821.3 XM_005262822.3 XM_005262823.3 XM_006714137.2 XM_017007884.1 XM_017007885.1 XM_017007886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4598190 c.491T>C p.I164T Chr.4: 186275792-186275792 on Build GRCh38
3392811 c.584T>C p.L195P Chr.4: 186275885-186275885 on Build GRCh38
1053942 c.537G>A p.T179T Chr.4: 186275838-186275838 on Build GRCh38
4331336 c.381G>A p.T127T Chr.4: 186275838-186275838 on Build GRCh38
1131163 c.595G>T p.A199S Chr.4: 186275896-186275896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766929913 Chr.4: 186275837-186275837 on Build GRCh38
rs764164418 Chr.4: 186275844-186275844 on Build GRCh38
rs574509278 Chr.4: 186275870-186275870 on Build GRCh38
rs371437683 Chr.4: 186275845-186275845 on Build GRCh38
rs748602158 Chr.4: 186275781-186275781 on Build GRCh38
rs768565290 Chr.4: 186275775-186275775 on Build GRCh38
rs752308147 Chr.4: 186275838-186275838 on Build GRCh38
rs368132889 Chr.4: 186275906-186275906 on Build GRCh38
rs763734062 Chr.4: 186275764-186275764 on Build GRCh38
rs534170853 Chr.4: 186275907-186275907 on Build GRCh38
rs142064053 Chr.4: 186275930-186275930 on Build GRCh38
rs374606272 Chr.4: 186275851-186275851 on Build GRCh38
rs781121860 Chr.4: 186275900-186275900 on Build GRCh38
rs1051927904 Chr.4: 186275917-186275917 on Build GRCh38
rs983294493 Chr.4: 186275817-186275817 on Build GRCh38
rs367559663 Chr.4: 186275808-186275808 on Build GRCh38
rs770391489 Chr.4: 186275784-186275784 on Build GRCh38
rs909066270 Chr.4: 186275827-186275827 on Build GRCh38
rs370991682 Chr.4: 186275777-186275777 on Build GRCh38
rs778213149 Chr.4: 186275913-186275913 on Build GRCh38
rs759231858 Chr.4: 186275831-186275831 on Build GRCh38
rs751669451 Chr.4: 186275853-186275853 on Build GRCh38
rs771805576 Chr.4: 186275936-186275936 on Build GRCh38
rs749817549 Chr.4: 186275915-186275915 on Build GRCh38
rs1057516695 Chr.4: 186275785-186275785 on Build GRCh38
rs374761594 Chr.4: 186275800-186275800 on Build GRCh38
rs959263144 Chr.4: 186275937-186275937 on Build GRCh38
rs190283743 Chr.4: 186275840-186275840 on Build GRCh38
rs747253590 Chr.4: 186275763-186275763 on Build GRCh38
rs933333847 Chr.4: 186275899-186275899 on Build GRCh38
rs759095254 Chr.4: 186275931-186275931 on Build GRCh38
rs771316448 Chr.4: 186275819-186275819 on Build GRCh38
rs773934995 Chr.4: 186275829-186275829 on Build GRCh38
rs771218036 Chr.4: 186275929-186275929 on Build GRCh38

View Full Product Details