Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110162105-110162364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A1
Entrez Gene ID
1282
Gene Name
collagen type IV alpha 1 chain
Gene Aliases
BSVD, RATOR
UniGene
Hs.17441
Gene Chromosome Location
Chr.13: 110148958-110307157 on Build GRCh38
Transcripts
NM_001845.5 XM_011521048.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4045913 c.4408G>A p.G1470R Chr.13: 110162284-110162284 on Build GRCh38
1248852 c.4459T>G p.L1487V Chr.13: 110162233-110162233 on Build GRCh38
3467103 c.4388G>A p.G1463E Chr.13: 110162304-110162304 on Build GRCh38
2262839 c.4441C>T p.R1481W Chr.13: 110162251-110162251 on Build GRCh38
356757 c.4366G>T p.D1456Y Chr.13: 110162326-110162326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747551299 Chr.13: 110162282-110162282 on Build GRCh38
rs137870576 Chr.13: 110162246-110162246 on Build GRCh38
rs369035639 Chr.13: 110162201-110162201 on Build GRCh38
rs199586038 Chr.13: 110162251-110162251 on Build GRCh38
rs781715366 Chr.13: 110162193-110162194 on Build GRCh38
rs767464071 Chr.13: 110162340-110162340 on Build GRCh38
rs751251728 Chr.13: 110162216-110162216 on Build GRCh38
rs761297535 Chr.13: 110162199-110162199 on Build GRCh38
rs776043824 Chr.13: 110162197-110162197 on Build GRCh38
rs200850535 Chr.13: 110162206-110162206 on Build GRCh38
rs765946790 Chr.13: 110162306-110162306 on Build GRCh38
rs779434104 Chr.13: 110162167-110162167 on Build GRCh38
rs567172177 Chr.13: 110162131-110162131 on Build GRCh38
rs201116557 Chr.13: 110162227-110162227 on Build GRCh38
rs957789748 Chr.13: 110162213-110162213 on Build GRCh38
rs772500580 Chr.13: 110162182-110162182 on Build GRCh38
rs867613837 Chr.13: 110162264-110162264 on Build GRCh38
rs752734207 Chr.13: 110162222-110162222 on Build GRCh38
rs754872910 Chr.13: 110162218-110162218 on Build GRCh38
rs759232482 Chr.13: 110162322-110162322 on Build GRCh38
rs775952266 Chr.13: 110162269-110162269 on Build GRCh38
rs772482135 Chr.13: 110162267-110162267 on Build GRCh38
rs535736541 Chr.13: 110162154-110162154 on Build GRCh38
rs11545878 Chr.13: 110162293-110162293 on Build GRCh38
rs746258731 Chr.13: 110162180-110162180 on Build GRCh38
rs767552075 Chr.13: 110162221-110162221 on Build GRCh38
rs777962282 Chr.13: 110162243-110162243 on Build GRCh38
rs149068853 Chr.13: 110162285-110162285 on Build GRCh38
rs1045694447 Chr.13: 110162126-110162126 on Build GRCh38
rs746214718 Chr.13: 110162266-110162266 on Build GRCh38
rs1004293546 Chr.13: 110162175-110162175 on Build GRCh38
rs376607450 Chr.13: 110162250-110162250 on Build GRCh38
rs977780860 Chr.13: 110162164-110162164 on Build GRCh38
rs370341214 Chr.13: 110162320-110162320 on Build GRCh38
rs772544207 Chr.13: 110162292-110162292 on Build GRCh38
rs748598865 Chr.13: 110162170-110162170 on Build GRCh38
rs768997641 Chr.13: 110162191-110162192 on Build GRCh38
rs748595524 Chr.13: 110162200-110162201 on Build GRCh38
rs185711350 Chr.13: 110162156-110162156 on Build GRCh38
rs887116296 Chr.13: 110162155-110162155 on Build GRCh38
rs1057518459 Chr.13: 110162304-110162304 on Build GRCh38
rs766074182 Chr.13: 110162209-110162209 on Build GRCh38

View Full Product Details