Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103476195-103476432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALHM3
Entrez Gene ID
119395
Gene Name
calcium homeostasis modulator 3
Gene Aliases
FAM26A, bA225H22.7
UniGene
Hs.364624
Gene Chromosome Location
Chr.10: 103472804-103479240 on Build GRCh38
Transcripts
NM_001129742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1199455 c.529C>T p.R177W Chr.10: 103476308-103476308 on Build GRCh38
5369999 c.477G>A p.E159E Chr.10: 103476360-103476360 on Build GRCh38
4497557 c.499C>T p.P167S Chr.10: 103476338-103476338 on Build GRCh38
5814521 c.440A>T p.Q147L Chr.10: 103476397-103476397 on Build GRCh38
1659397 c.447G>A p.Q149Q Chr.10: 103476390-103476390 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909025067 Chr.10: 103476368-103476368 on Build GRCh38
rs757750026 Chr.10: 103476323-103476323 on Build GRCh38
rs1005642755 Chr.10: 103476265-103476265 on Build GRCh38
rs149475857 Chr.10: 103476351-103476351 on Build GRCh38
rs751696710 Chr.10: 103476359-103476359 on Build GRCh38
rs958909925 Chr.10: 103476218-103476218 on Build GRCh38
rs189405481 Chr.10: 103476308-103476308 on Build GRCh38
rs574408741 Chr.10: 103476264-103476264 on Build GRCh38
rs752667505 Chr.10: 103476300-103476300 on Build GRCh38
rs762179943 Chr.10: 103476285-103476285 on Build GRCh38
rs965747946 Chr.10: 103476287-103476287 on Build GRCh38
rs756327320 Chr.10: 103476283-103476283 on Build GRCh38
rs768376244 Chr.10: 103476376-103476376 on Build GRCh38
rs964720221 Chr.10: 103476383-103476383 on Build GRCh38
rs776938886 Chr.10: 103476278-103476278 on Build GRCh38
rs773926654 Chr.10: 103476407-103476407 on Build GRCh38
rs764966146 Chr.10: 103476244-103476244 on Build GRCh38
rs780051327 Chr.10: 103476342-103476342 on Build GRCh38
rs765329602 Chr.10: 103476272-103476272 on Build GRCh38
rs199912946 Chr.10: 103476307-103476307 on Build GRCh38
rs372235948 Chr.10: 103476271-103476271 on Build GRCh38
rs1021720393 Chr.10: 103476245-103476245 on Build GRCh38
rs755160395 Chr.10: 103476260-103476260 on Build GRCh38
rs528807632 Chr.10: 103476336-103476336 on Build GRCh38
rs758953571 Chr.10: 103476306-103476306 on Build GRCh38
rs867561113 Chr.10: 103476327-103476327 on Build GRCh38
rs895729541 Chr.10: 103476239-103476239 on Build GRCh38

View Full Product Details