Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184341631-184341904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM131A
Entrez Gene ID
131408
Gene Name
family with sequence similarity 131 member A
Gene Aliases
C3orf40, FLAT715, PRO1378
UniGene
Hs.591307
Gene Chromosome Location
Chr.3: 184335924-184346275 on Build GRCh38
Transcripts
NM_001171093.1 NM_144635.4 XM_005247113.2 XM_005247114.3 XM_011512413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1617270 c.32C>T p.P11L Chr.3: 184341865-184341865 on Build GRCh38
325296 c.278C>T p.A93V Chr.3: 184341770-184341770 on Build GRCh38
3331252 c.254C>T p.A85V Chr.3: 184341746-184341746 on Build GRCh38
6289873 c.325+9C>T p.? Chr.3: 184341826-184341826 on Build GRCh38
5455170 c.311G>T p.R104M Chr.3: 184341803-184341803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028053042 Chr.3: 184341791-184341791 on Build GRCh38
rs563236472 Chr.3: 184341680-184341680 on Build GRCh38
rs549519004 Chr.3: 184341829-184341829 on Build GRCh38
rs187325668 Chr.3: 184341794-184341794 on Build GRCh38
rs373794061 Chr.3: 184341866-184341866 on Build GRCh38
rs753120304 Chr.3: 184341865-184341865 on Build GRCh38
rs995240932 Chr.3: 184341702-184341702 on Build GRCh38
rs760547288 Chr.3: 184341790-184341790 on Build GRCh38
rs765358430 Chr.3: 184341718-184341718 on Build GRCh38
rs573329942 Chr.3: 184341655-184341655 on Build GRCh38
rs781565540 Chr.3: 184341851-184341851 on Build GRCh38
rs149602757 Chr.3: 184341668-184341668 on Build GRCh38
rs937821012 Chr.3: 184341665-184341665 on Build GRCh38
rs758581063 Chr.3: 184341731-184341731 on Build GRCh38
rs750456077 Chr.3: 184341805-184341805 on Build GRCh38
rs780347486 Chr.3: 184341732-184341732 on Build GRCh38
rs376762321 Chr.3: 184341789-184341789 on Build GRCh38
rs748027446 Chr.3: 184341839-184341839 on Build GRCh38
rs767851624 Chr.3: 184341874-184341874 on Build GRCh38
rs770440803 Chr.3: 184341747-184341747 on Build GRCh38
rs761876228 Chr.3: 184341714-184341714 on Build GRCh38
rs371516806 Chr.3: 184341864-184341864 on Build GRCh38
rs755411212 Chr.3: 184341742-184341742 on Build GRCh38
rs545636303 Chr.3: 184341745-184341745 on Build GRCh38
rs367965687 Chr.3: 184341834-184341834 on Build GRCh38
rs561344087 Chr.3: 184341832-184341832 on Build GRCh38
rs1032534707 Chr.3: 184341845-184341845 on Build GRCh38
rs888344450 Chr.3: 184341730-184341730 on Build GRCh38
rs755289531 Chr.3: 184341835-184341835 on Build GRCh38
rs770437990 Chr.3: 184341763-184341763 on Build GRCh38
rs754948255 Chr.3: 184341686-184341686 on Build GRCh38
rs772990515 Chr.3: 184341684-184341684 on Build GRCh38
rs560529445 Chr.3: 184341746-184341746 on Build GRCh38
rs750513043 Chr.3: 184341721-184341721 on Build GRCh38
rs767332266 Chr.3: 184341697-184341697 on Build GRCh38
rs770624246 Chr.3: 184341691-184341691 on Build GRCh38
rs777353879 Chr.3: 184341756-184341756 on Build GRCh38
rs1033386821 Chr.3: 184341873-184341873 on Build GRCh38
rs748835883 Chr.3: 184341761-184341761 on Build GRCh38
rs771969220 Chr.3: 184341777-184341777 on Build GRCh38
rs578184852 Chr.3: 184341683-184341683 on Build GRCh38
rs759395407 Chr.3: 184341694-184341694 on Build GRCh38
rs759216346 Chr.3: 184341768-184341768 on Build GRCh38
rs900545415 Chr.3: 184341687-184341687 on Build GRCh38
rs375404910 Chr.3: 184341679-184341679 on Build GRCh38
rs774098952 Chr.3: 184341767-184341767 on Build GRCh38
rs752716745 Chr.3: 184341705-184341705 on Build GRCh38
rs751848994 Chr.3: 184341740-184341740 on Build GRCh38
rs747631634 Chr.3: 184341681-184341681 on Build GRCh38
rs1026934854 Chr.3: 184341706-184341706 on Build GRCh38

View Full Product Details