Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 38481147-38481390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM98B
Entrez Gene ID
283742
Gene Name
family with sequence similarity 98 member B
Gene Aliases
-
UniGene
Hs.625289
Gene Chromosome Location
Chr.15: 38454127-38487710 on Build GRCh38
Transcripts
NM_173611.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2268672 c.744T>C p.D248D Chr.15: 38481306-38481306 on Build GRCh38
3420259 c.795G>A p.T265T Chr.15: 38481357-38481357 on Build GRCh38
2268669 c.731T>G p.V244G Chr.15: 38481293-38481293 on Build GRCh38
2268671 c.744T>C p.D248D Chr.15: 38481306-38481306 on Build GRCh38
960993 c.769C>T p.R257C Chr.15: 38481331-38481331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751904798 Chr.15: 38481329-38481329 on Build GRCh38
rs756841894 Chr.15: 38481213-38481213 on Build GRCh38
rs775161500 Chr.15: 38481241-38481241 on Build GRCh38
rs373769983 Chr.15: 38481283-38481283 on Build GRCh38
rs897644656 Chr.15: 38481203-38481207 on Build GRCh38
rs780403041 Chr.15: 38481317-38481317 on Build GRCh38
rs1014117588 Chr.15: 38481321-38481321 on Build GRCh38
rs774585657 Chr.15: 38481355-38481355 on Build GRCh38
rs199750152 Chr.15: 38481293-38481293 on Build GRCh38
rs549269634 Chr.15: 38481181-38481181 on Build GRCh38
rs755739118 Chr.15: 38481335-38481335 on Build GRCh38
rs778623343 Chr.15: 38481360-38481360 on Build GRCh38
rs775630121 Chr.15: 38481277-38481277 on Build GRCh38
rs1036728448 Chr.15: 38481194-38481194 on Build GRCh38
rs761851653 Chr.15: 38481270-38481270 on Build GRCh38
rs1032366853 Chr.15: 38481364-38481364 on Build GRCh38
rs376754001 Chr.15: 38481336-38481336 on Build GRCh38
rs542841170 Chr.15: 38481260-38481260 on Build GRCh38
rs760884102 Chr.15: 38481256-38481256 on Build GRCh38
rs758922386 Chr.15: 38481313-38481313 on Build GRCh38
rs768788629 Chr.15: 38481332-38481332 on Build GRCh38
rs941952035 Chr.15: 38481179-38481179 on Build GRCh38
rs1001254343 Chr.15: 38481222-38481222 on Build GRCh38
rs746255605 Chr.15: 38481195-38481195 on Build GRCh38
rs761793572 Chr.15: 38481357-38481357 on Build GRCh38
rs1007938460 Chr.15: 38481285-38481285 on Build GRCh38

View Full Product Details