Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53713662-53713931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCOCO1
Entrez Gene ID
57658
Gene Name
calcium binding and coiled-coil domain 1
Gene Aliases
Cocoa, PP13275, calphoglin
UniGene
Hs.156667
Gene Chromosome Location
Chr.12: 53711118-53727523 on Build GRCh38
Transcripts
NM_001143682.1 NM_020898.2 NR_026554.1 XM_011538601.1 XM_011538602.1 XM_011538603.1 XM_011538604.1 XM_017019707.1 XM_017019708.1 XM_017019709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3462726 c.1651C>T p.P551S Chr.12: 53713841-53713841 on Build GRCh38
4782839 c.1549C>T p.P517S Chr.12: 53713757-53713757 on Build GRCh38
3462725 c.1699C>T p.P567S Chr.12: 53713793-53713793 on Build GRCh38
4899452 c.1557C>T p.P519P Chr.12: 53713749-53713749 on Build GRCh38
3462724 c.1743C>T p.P581P Chr.12: 53713749-53713749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755287694 Chr.12: 53713716-53713716 on Build GRCh38
rs753405269 Chr.12: 53713873-53713878 on Build GRCh38
rs757191074 Chr.12: 53713802-53713802 on Build GRCh38
rs767996532 Chr.12: 53713822-53713823 on Build GRCh38
rs747381370 Chr.12: 53713837-53713837 on Build GRCh38
rs774453792 Chr.12: 53713817-53713817 on Build GRCh38
rs868841389 Chr.12: 53713724-53713724 on Build GRCh38
rs747400206 Chr.12: 53713897-53713897 on Build GRCh38
rs776903582 Chr.12: 53713742-53713742 on Build GRCh38
rs377156371 Chr.12: 53713880-53713880 on Build GRCh38
rs34229062 Chr.12: 53713810-53713810 on Build GRCh38
rs761049308 Chr.12: 53713727-53713727 on Build GRCh38
rs780632107 Chr.12: 53713683-53713683 on Build GRCh38
rs779502814 Chr.12: 53713888-53713888 on Build GRCh38
rs751535599 Chr.12: 53713798-53713798 on Build GRCh38
rs763755443 Chr.12: 53713789-53713789 on Build GRCh38
rs202198086 Chr.12: 53713815-53713815 on Build GRCh38
rs752956910 Chr.12: 53713780-53713780 on Build GRCh38
rs144266090 Chr.12: 53713725-53713725 on Build GRCh38
rs878988874 Chr.12: 53713847-53713847 on Build GRCh38
rs531381809 Chr.12: 53713853-53713853 on Build GRCh38
rs200862370 Chr.12: 53713701-53713701 on Build GRCh38
rs758227615 Chr.12: 53713786-53713786 on Build GRCh38
rs766506396 Chr.12: 53713738-53713738 on Build GRCh38
rs769523405 Chr.12: 53713836-53713836 on Build GRCh38
rs759749697 Chr.12: 53713849-53713849 on Build GRCh38
rs759658749 Chr.12: 53713755-53713755 on Build GRCh38
rs200116362 Chr.12: 53713902-53713902 on Build GRCh38
rs745358997 Chr.12: 53713699-53713699 on Build GRCh38
rs934031692 Chr.12: 53713900-53713900 on Build GRCh38
rs560882432 Chr.12: 53713840-53713840 on Build GRCh38
rs941833602 Chr.12: 53713864-53713864 on Build GRCh38
rs868636620 Chr.12: 53713790-53713790 on Build GRCh38
rs975706877 Chr.12: 53713812-53713812 on Build GRCh38
rs763034547 Chr.12: 53713868-53713868 on Build GRCh38
rs369598454 Chr.12: 53713708-53713708 on Build GRCh38
rs116674128 Chr.12: 53713813-53713813 on Build GRCh38
rs778600659 Chr.12: 53713896-53713896 on Build GRCh38
rs749318044 Chr.12: 53713807-53713807 on Build GRCh38
rs761660061 Chr.12: 53713874-53713874 on Build GRCh38
rs781768317 Chr.12: 53713906-53713906 on Build GRCh38
rs772520577 Chr.12: 53713823-53713823 on Build GRCh38
rs372844638 Chr.12: 53713904-53713904 on Build GRCh38
rs200742427 Chr.12: 53713884-53713884 on Build GRCh38
rs777273475 Chr.12: 53713844-53713844 on Build GRCh38
rs369102896 Chr.12: 53713894-53713894 on Build GRCh38
rs748601288 Chr.12: 53713714-53713714 on Build GRCh38
rs769992886 Chr.12: 53713704-53713704 on Build GRCh38
rs765623025 Chr.12: 53713756-53713756 on Build GRCh38
rs748822880 Chr.12: 53713822-53713822 on Build GRCh38
rs140431811 Chr.12: 53713878-53713878 on Build GRCh38
rs369656859 Chr.12: 53713869-53713869 on Build GRCh38
rs753554149 Chr.12: 53713893-53713893 on Build GRCh38
rs199692489 Chr.12: 53713719-53713719 on Build GRCh38
rs751349975 Chr.12: 53713871-53713871 on Build GRCh38

View Full Product Details