Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41215700-41215933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXD1
Entrez Gene ID
161829
Gene Name
exonuclease 3'-5' domain containing 1
Gene Aliases
EXDL1
UniGene
Hs.307999
Gene Chromosome Location
Chr.15: 41182728-41231074 on Build GRCh38
Transcripts
NM_001286441.1 NM_152596.3 XM_006720415.3 XM_011521298.2 XM_011521299.2 XM_011521300.2 XM_011521301.2 XM_017021970.1 XM_017021971.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1608338 c.273+4A>T p.? Chr.15: 41215771-41215771 on Build GRCh38
5451351 c.230C>T p.T77I Chr.15: 41215818-41215818 on Build GRCh38
1205787 c.269C>T p.A90V Chr.15: 41215779-41215779 on Build GRCh38
4418132 c.215-6T>C p.? Chr.15: 41215839-41215839 on Build GRCh38
7197745 c.264T>A p.F88L Chr.15: 41215784-41215784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778348080 Chr.15: 41215792-41215792 on Build GRCh38
rs753514853 Chr.15: 41215863-41215863 on Build GRCh38
rs763820609 Chr.15: 41215776-41215776 on Build GRCh38
rs763620283 Chr.15: 41215847-41215847 on Build GRCh38
rs182024066 Chr.15: 41215862-41215862 on Build GRCh38
rs771456397 Chr.15: 41215851-41215851 on Build GRCh38
rs535853801 Chr.15: 41215815-41215815 on Build GRCh38
rs1032278719 Chr.15: 41215886-41215886 on Build GRCh38
rs575535440 Chr.15: 41215883-41215883 on Build GRCh38
rs750530986 Chr.15: 41215739-41215739 on Build GRCh38
rs983590639 Chr.15: 41215852-41215852 on Build GRCh38
rs766693741 Chr.15: 41215758-41215758 on Build GRCh38
rs1011869244 Chr.15: 41215902-41215902 on Build GRCh38
rs567978492 Chr.15: 41215901-41215901 on Build GRCh38
rs745859150 Chr.15: 41215793-41215793 on Build GRCh38
rs761435901 Chr.15: 41215871-41215871 on Build GRCh38
rs770041221 Chr.15: 41215846-41215846 on Build GRCh38
rs1027888703 Chr.15: 41215817-41215817 on Build GRCh38
rs373678111 Chr.15: 41215743-41215743 on Build GRCh38
rs376055755 Chr.15: 41215780-41215780 on Build GRCh38
rs746829239 Chr.15: 41215813-41215813 on Build GRCh38
rs376298586 Chr.15: 41215887-41215887 on Build GRCh38
rs1021965757 Chr.15: 41215904-41215904 on Build GRCh38
rs142913906 Chr.15: 41215799-41215799 on Build GRCh38
rs144119621 Chr.15: 41215804-41215804 on Build GRCh38
rs199630693 Chr.15: 41215860-41215860 on Build GRCh38
rs371053623 Chr.15: 41215790-41215790 on Build GRCh38
rs1031410012 Chr.15: 41215752-41215752 on Build GRCh38
rs751948603 Chr.15: 41215766-41215766 on Build GRCh38
rs572574688 Chr.15: 41215856-41215856 on Build GRCh38
rs776940670 Chr.15: 41215830-41215830 on Build GRCh38
rs368921977 Chr.15: 41215770-41215770 on Build GRCh38
rs8025916 Chr.15: 41215839-41215839 on Build GRCh38
rs201768915 Chr.15: 41215779-41215779 on Build GRCh38
rs774662909 Chr.15: 41215855-41215855 on Build GRCh38
rs373412692 Chr.15: 41215884-41215884 on Build GRCh38
rs1018995284 Chr.15: 41215775-41215775 on Build GRCh38

View Full Product Details