Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 64249469-64249730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESR2
Entrez Gene ID
2100
Gene Name
estrogen receptor 2
Gene Aliases
ER-BETA, ESR-BETA, ESRB, ESTRB, Erb, NR3A2
UniGene
Hs.660607
Gene Chromosome Location
Chr.14: 64226712-64338631 on Build GRCh38
Transcripts
NM_001040275.1 NM_001214902.1 NM_001271876.1 NM_001271877.1 NM_001291712.1 NM_001291723.1 NM_001437.2 NR_073496.1 NR_073497.1 XM_017021079.1 XM_017021080.1 XM_017021081.1 XM_017021082.1 XM_017021083.1 XM_017021084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419956 c.1186G>T p.E396* Chr.14: 64249585-64249585 on Build GRCh38
6204558 c.1190A>G p.Y397C Chr.14: 64249581-64249581 on Build GRCh38
5421166 c.1194C>G p.L398L Chr.14: 64249577-64249577 on Build GRCh38
5421162 c.1194C>G p.L398L Chr.14: 64249577-64249577 on Build GRCh38
3744410 c.1205C>T p.A402V Chr.14: 64249566-64249566 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1256054 Chr.14: 64249595-64249595 on Build GRCh38
rs776592617 Chr.14: 64249680-64249680 on Build GRCh38
rs776023335 Chr.14: 64249702-64249702 on Build GRCh38
rs768030327 Chr.14: 64249616-64249616 on Build GRCh38
rs374768593 Chr.14: 64249631-64249631 on Build GRCh38
rs186667034 Chr.14: 64249520-64249520 on Build GRCh38
rs144249047 Chr.14: 64249669-64249669 on Build GRCh38
rs112017626 Chr.14: 64249629-64249629 on Build GRCh38
rs141474553 Chr.14: 64249679-64249679 on Build GRCh38
rs1057398397 Chr.14: 64249531-64249531 on Build GRCh38
rs778031158 Chr.14: 64249552-64249552 on Build GRCh38
rs762432960 Chr.14: 64249695-64249695 on Build GRCh38
rs753050523 Chr.14: 64249620-64249620 on Build GRCh38
rs759226708 Chr.14: 64249509-64249509 on Build GRCh38
rs772723029 Chr.14: 64249693-64249693 on Build GRCh38
rs1012693115 Chr.14: 64249582-64249582 on Build GRCh38
rs771907276 Chr.14: 64249564-64249564 on Build GRCh38
rs766765307 Chr.14: 64249522-64249522 on Build GRCh38
rs753066940 Chr.14: 64249547-64249547 on Build GRCh38
rs776464052 Chr.14: 64249591-64249591 on Build GRCh38
rs369437676 Chr.14: 64249524-64249524 on Build GRCh38
rs760024678 Chr.14: 64249613-64249613 on Build GRCh38
rs756507443 Chr.14: 64249621-64249621 on Build GRCh38
rs745947456 Chr.14: 64249663-64249663 on Build GRCh38
rs775455013 Chr.14: 64249512-64249512 on Build GRCh38
rs201833437 Chr.14: 64249697-64249697 on Build GRCh38
rs111471356 Chr.14: 64249554-64249554 on Build GRCh38
rs34373341 Chr.14: 64249705-64249706 on Build GRCh38
rs77486742 Chr.14: 64249692-64249692 on Build GRCh38
rs772629994 Chr.14: 64249609-64249609 on Build GRCh38
rs1010629502 Chr.14: 64249615-64249615 on Build GRCh38
rs927692826 Chr.14: 64249587-64249587 on Build GRCh38
rs1004400157 Chr.14: 64249652-64249652 on Build GRCh38
rs79216673 Chr.14: 64249562-64249562 on Build GRCh38
rs888179672 Chr.14: 64249633-64249633 on Build GRCh38
rs764756707 Chr.14: 64249608-64249608 on Build GRCh38
rs986297029 Chr.14: 64249694-64249694 on Build GRCh38
rs774382661 Chr.14: 64249501-64249501 on Build GRCh38
rs764274252 Chr.14: 64249623-64249623 on Build GRCh38
rs45624541 Chr.14: 64249583-64249583 on Build GRCh38
rs78255744 Chr.14: 64249548-64249548 on Build GRCh38
rs755463625 Chr.14: 64249529-64249529 on Build GRCh38
rs376348920 Chr.14: 64249637-64249637 on Build GRCh38
rs373170989 Chr.14: 64249555-64249555 on Build GRCh38
rs531227990 Chr.14: 64249643-64249643 on Build GRCh38
rs80118085 Chr.14: 64249664-64249664 on Build GRCh38
rs550916443 Chr.14: 64249511-64249511 on Build GRCh38
rs781583718 Chr.14: 64249532-64249532 on Build GRCh38
rs778982958 Chr.14: 64249650-64249650 on Build GRCh38
rs72548752 Chr.14: 64249565-64249565 on Build GRCh38
rs546290724 Chr.14: 64249513-64249513 on Build GRCh38
rs147847668 Chr.14: 64249670-64249670 on Build GRCh38
rs754008155 Chr.14: 64249636-64249636 on Build GRCh38
rs747797582 Chr.14: 64249691-64249691 on Build GRCh38
rs775365033 Chr.14: 64249571-64249571 on Build GRCh38
rs1030555125 Chr.14: 64249675-64249675 on Build GRCh38
rs778878210 Chr.14: 64249556-64249556 on Build GRCh38
rs746812518 Chr.14: 64249564-64249565 on Build GRCh38
rs768417560 Chr.14: 64249589-64249589 on Build GRCh38

View Full Product Details