Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 116988592-116988865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW8
Entrez Gene ID
26259
Gene Name
F-box and WD repeat domain containing 8
Gene Aliases
FBW6, FBW8, FBX29, FBXO29, FBXW6
UniGene
Hs.624537
Gene Chromosome Location
Chr.12: 116910949-117032498 on Build GRCh38
Transcripts
NM_012174.1 NM_153348.2 XM_005253867.2 XM_005253868.3 XM_017019175.1 XM_017019176.1 XM_017019177.1 XM_017019178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6135686 c.1142T>G p.L381R Chr.12: 116988772-116988772 on Build GRCh38
5612674 c.1153G>A p.G385S Chr.12: 116988783-116988783 on Build GRCh38
4826500 c.1174G>A p.D392N Chr.12: 116988804-116988804 on Build GRCh38
935586 c.1053T>C p.V351V Chr.12: 116988683-116988683 on Build GRCh38
935587 c.1117G>A p.A373T Chr.12: 116988747-116988747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761811567 Chr.12: 116988657-116988657 on Build GRCh38
rs777128618 Chr.12: 116988654-116988654 on Build GRCh38
rs751296718 Chr.12: 116988711-116988711 on Build GRCh38
rs752616383 Chr.12: 116988782-116988782 on Build GRCh38
rs774578340 Chr.12: 116988775-116988775 on Build GRCh38
rs763233036 Chr.12: 116988669-116988669 on Build GRCh38
rs148779892 Chr.12: 116988665-116988665 on Build GRCh38
rs199846352 Chr.12: 116988777-116988777 on Build GRCh38
rs1007869326 Chr.12: 116988698-116988698 on Build GRCh38
rs778548510 Chr.12: 116988798-116988798 on Build GRCh38
rs774812881 Chr.12: 116988763-116988763 on Build GRCh38
rs760584211 Chr.12: 116988783-116988783 on Build GRCh38
rs761904700 Chr.12: 116988714-116988714 on Build GRCh38
rs1030384886 Chr.12: 116988737-116988737 on Build GRCh38
rs771285128 Chr.12: 116988774-116988774 on Build GRCh38
rs766323509 Chr.12: 116988705-116988705 on Build GRCh38
rs759275514 Chr.12: 116988776-116988776 on Build GRCh38
rs756891269 Chr.12: 116988721-116988721 on Build GRCh38
rs759311689 Chr.12: 116988714-116988714 on Build GRCh38
rs371693667 Chr.12: 116988668-116988668 on Build GRCh38
rs763429979 Chr.12: 116988774-116988775 on Build GRCh38
rs368889689 Chr.12: 116988749-116988749 on Build GRCh38
rs747611900 Chr.12: 116988643-116988643 on Build GRCh38
rs151338590 Chr.12: 116988759-116988759 on Build GRCh38
rs753539482 Chr.12: 116988720-116988720 on Build GRCh38
rs17853730 Chr.12: 116988768-116988768 on Build GRCh38
rs547950465 Chr.12: 116988787-116988787 on Build GRCh38
rs780466464 Chr.12: 116988733-116988733 on Build GRCh38
rs781570701 Chr.12: 116988750-116988750 on Build GRCh38
rs375481391 Chr.12: 116988788-116988788 on Build GRCh38
rs748900112 Chr.12: 116988627-116988627 on Build GRCh38
rs765262152 Chr.12: 116988739-116988740 on Build GRCh38
rs142445679 Chr.12: 116988716-116988716 on Build GRCh38
rs772466610 Chr.12: 116988639-116988639 on Build GRCh38
rs781585590 Chr.12: 116988811-116988811 on Build GRCh38
rs778063449 Chr.12: 116988830-116988830 on Build GRCh38
rs769037612 Chr.12: 116988648-116988648 on Build GRCh38
rs772659476 Chr.12: 116988806-116988806 on Build GRCh38
rs953521136 Chr.12: 116988743-116988743 on Build GRCh38
rs750970417 Chr.12: 116988748-116988748 on Build GRCh38
rs755615235 Chr.12: 116988717-116988717 on Build GRCh38
rs374795754 Chr.12: 116988741-116988741 on Build GRCh38
rs748649049 Chr.12: 116988817-116988817 on Build GRCh38
rs763321710 Chr.12: 116988765-116988765 on Build GRCh38
rs755595921 Chr.12: 116988620-116988620 on Build GRCh38
rs777430223 Chr.12: 116988625-116988625 on Build GRCh38
rs749394927 Chr.12: 116988837-116988837 on Build GRCh38
rs756687387 Chr.12: 116988797-116988797 on Build GRCh38
rs140698430 Chr.12: 116988784-116988784 on Build GRCh38
rs775811121 Chr.12: 116988641-116988641 on Build GRCh38
rs771338638 Chr.12: 116988758-116988758 on Build GRCh38
rs755328748 Chr.12: 116988807-116988807 on Build GRCh38
rs369751629 Chr.12: 116988822-116988822 on Build GRCh38

View Full Product Details