Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38232988-38233255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDHD2
Entrez Gene ID
23259
Gene Name
DDHD domain containing 2
Gene Aliases
SAMWD1, SPG54, iPLA(1)gamma
UniGene
Hs.434966
Gene Chromosome Location
Chr.8: 38231343-38273442 on Build GRCh38
Transcripts
NM_001164232.1 NM_001164234.1 NM_015214.2 XM_005273454.2 XM_005273455.3 XM_005273456.3 XM_011544456.1 XM_017013252.1 XM_017013253.1 XM_017013254.1 XM_017013255.1 XM_017013256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203189 c.74C>A p.S25Y Chr.8: 38233068-38233068 on Build GRCh38
5485016 c.137A>G p.Y46C Chr.8: 38233131-38233131 on Build GRCh38
6516099 c.5C>A p.S2* Chr.8: 38232999-38232999 on Build GRCh38
2960171 c.116C>A p.P39Q Chr.8: 38233110-38233110 on Build GRCh38
422136 c.202G>A p.E68K Chr.8: 38233196-38233196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143549962 Chr.8: 38233047-38233047 on Build GRCh38
rs773256513 Chr.8: 38233209-38233209 on Build GRCh38
rs771376379 Chr.8: 38233011-38233011 on Build GRCh38
rs766240465 Chr.8: 38233068-38233068 on Build GRCh38
rs932523071 Chr.8: 38233123-38233123 on Build GRCh38
rs112061720 Chr.8: 38233116-38233116 on Build GRCh38
rs780610008 Chr.8: 38233084-38233084 on Build GRCh38
rs779390449 Chr.8: 38233155-38233155 on Build GRCh38
rs768909651 Chr.8: 38233183-38233183 on Build GRCh38
rs201334641 Chr.8: 38233013-38233013 on Build GRCh38
rs761777370 Chr.8: 38233039-38233039 on Build GRCh38
rs778229764 Chr.8: 38233126-38233126 on Build GRCh38
rs200883537 Chr.8: 38233133-38233133 on Build GRCh38
rs762638689 Chr.8: 38233066-38233066 on Build GRCh38
rs775728459 Chr.8: 38233038-38233038 on Build GRCh38
rs750194962 Chr.8: 38233048-38233048 on Build GRCh38
rs754719646 Chr.8: 38233078-38233078 on Build GRCh38
rs771194032 Chr.8: 38233145-38233145 on Build GRCh38
rs746020888 Chr.8: 38233159-38233159 on Build GRCh38
rs756560473 Chr.8: 38233110-38233110 on Build GRCh38
rs145837688 Chr.8: 38233108-38233108 on Build GRCh38
rs903117028 Chr.8: 38233118-38233118 on Build GRCh38
rs774404330 Chr.8: 38233012-38233012 on Build GRCh38
rs751170158 Chr.8: 38233075-38233075 on Build GRCh38
rs138068744 Chr.8: 38233033-38233033 on Build GRCh38
rs1009555066 Chr.8: 38233090-38233090 on Build GRCh38
rs763017121 Chr.8: 38233228-38233228 on Build GRCh38
rs182341575 Chr.8: 38233015-38233015 on Build GRCh38
rs760860946 Chr.8: 38233175-38233175 on Build GRCh38
rs370564627 Chr.8: 38233164-38233164 on Build GRCh38

View Full Product Details