Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68919715-68919984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX50
Entrez Gene ID
79009
Gene Name
DEAD-box helicase 50
Gene Aliases
GU2, GUB, RH-II/GuB, mcdrh
UniGene
Hs.522984
Gene Chromosome Location
Chr.10: 68901278-68956312 on Build GRCh38
Transcripts
NM_024045.1 XM_005270148.2 XM_011540144.2 XM_017016626.1 XM_017016627.1 XM_017016628.1 XM_017016629.1 XM_017016630.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6020835 c.1119G>C p.Q373H Chr.10: 68919861-68919861 on Build GRCh38
4015334 c.1136G>T p.G379V Chr.10: 68919878-68919878 on Build GRCh38
3439938 c.1101C>T p.I367I Chr.10: 68919843-68919843 on Build GRCh38
6316937 c.1095G>T p.L365F Chr.10: 68919837-68919837 on Build GRCh38
3782551 c.1189G>A p.E397K Chr.10: 68919931-68919931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770880483 Chr.10: 68919847-68919847 on Build GRCh38
rs957398521 Chr.10: 68919825-68919825 on Build GRCh38
rs767242175 Chr.10: 68919875-68919875 on Build GRCh38
rs762917933 Chr.10: 68919846-68919846 on Build GRCh38
rs575007030 Chr.10: 68919881-68919881 on Build GRCh38
rs200542425 Chr.10: 68919814-68919814 on Build GRCh38
rs201478746 Chr.10: 68919840-68919840 on Build GRCh38
rs368897102 Chr.10: 68919822-68919822 on Build GRCh38
rs538366988 Chr.10: 68919899-68919899 on Build GRCh38
rs527480474 Chr.10: 68919768-68919768 on Build GRCh38
rs577377946 Chr.10: 68919894-68919894 on Build GRCh38
rs117029940 Chr.10: 68919756-68919756 on Build GRCh38
rs759386831 Chr.10: 68919874-68919874 on Build GRCh38
rs967602258 Chr.10: 68919929-68919929 on Build GRCh38
rs372290555 Chr.10: 68919805-68919805 on Build GRCh38
rs913159602 Chr.10: 68919885-68919885 on Build GRCh38
rs763451791 Chr.10: 68919903-68919903 on Build GRCh38
rs779178417 Chr.10: 68919783-68919783 on Build GRCh38
rs754343476 Chr.10: 68919930-68919930 on Build GRCh38
rs760288224 Chr.10: 68919892-68919892 on Build GRCh38
rs981262213 Chr.10: 68919938-68919938 on Build GRCh38
rs772232150 Chr.10: 68919799-68919799 on Build GRCh38
rs757717045 Chr.10: 68919942-68919942 on Build GRCh38
rs774242439 Chr.10: 68919852-68919852 on Build GRCh38
rs371253759 Chr.10: 68919889-68919889 on Build GRCh38
rs146773324 Chr.10: 68919798-68919798 on Build GRCh38

View Full Product Details