Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 71570737-71570974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
UniGene
Hs.656032
Gene Chromosome Location
Chr.10: 71396934-71815947 on Build GRCh38
Transcripts
NM_001171930.1 NM_001171931.1 NM_001171932.1 NM_022124.5 NM_052836.3 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_017016499.1 XM_017016500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5371065 c.881C>T p.S294F Chr.10: 71570911-71570911 on Build GRCh38
6885511 c.728C>T p.T243I Chr.10: 71570893-71570893 on Build GRCh38
4504066 c.670C>T p.L224L Chr.10: 71570820-71570820 on Build GRCh38
3440198 c.712G>A p.D238N Chr.10: 71570862-71570862 on Build GRCh38
5371062 c.746C>T p.S249F Chr.10: 71570911-71570911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752101206 Chr.10: 71570864-71570864 on Build GRCh38
rs768084499 Chr.10: 71570861-71570861 on Build GRCh38
rs766721575 Chr.10: 71570827-71570827 on Build GRCh38
rs751987327 Chr.10: 71570832-71570832 on Build GRCh38
rs755616454 Chr.10: 71570868-71570868 on Build GRCh38
rs530434456 Chr.10: 71570865-71570865 on Build GRCh38
rs551858055 Chr.10: 71570860-71570860 on Build GRCh38
rs1041759385 Chr.10: 71570867-71570867 on Build GRCh38
rs754239533 Chr.10: 71570823-71570823 on Build GRCh38
rs779733078 Chr.10: 71570775-71570775 on Build GRCh38
rs749507744 Chr.10: 71570948-71570948 on Build GRCh38
rs773456103 Chr.10: 71570807-71570807 on Build GRCh38
rs142846689 Chr.10: 71570759-71570759 on Build GRCh38
rs538068566 Chr.10: 71570897-71570897 on Build GRCh38
rs766217256 Chr.10: 71570906-71570906 on Build GRCh38
rs745791414 Chr.10: 71570779-71570779 on Build GRCh38
rs769896655 Chr.10: 71570918-71570918 on Build GRCh38
rs761972494 Chr.10: 71570783-71570783 on Build GRCh38
rs763214670 Chr.10: 71570810-71570810 on Build GRCh38
rs757037273 Chr.10: 71570895-71570895 on Build GRCh38
rs753526835 Chr.10: 71570893-71570893 on Build GRCh38
rs121908354 Chr.10: 71570884-71570884 on Build GRCh38
rs746961144 Chr.10: 71570917-71570917 on Build GRCh38
rs745668474 Chr.10: 71570903-71570903 on Build GRCh38
rs773224102 Chr.10: 71570937-71570937 on Build GRCh38
rs772135580 Chr.10: 71570780-71570780 on Build GRCh38
rs764515216 Chr.10: 71570798-71570798 on Build GRCh38
rs548239921 Chr.10: 71570951-71570951 on Build GRCh38
rs780068885 Chr.10: 71570913-71570913 on Build GRCh38

View Full Product Details