Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 694537-694778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL16
Entrez Gene ID
146330
Gene Name
F-box and leucine rich repeat protein 16
Gene Aliases
C16orf22, Fbl16, c380A1.1
UniGene
Hs.513244
Gene Chromosome Location
Chr.16: 692500-705825 on Build GRCh38
Transcripts
NM_153350.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5851471 c.1250A>C p.H417P Chr.16: 694675-694675 on Build GRCh38
5524958 c.1266G>A p.G422G Chr.16: 694659-694659 on Build GRCh38
4062760 c.1239C>G p.F413L Chr.16: 694686-694686 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756113309 Chr.16: 694685-694685 on Build GRCh38
rs201039799 Chr.16: 694606-694606 on Build GRCh38
rs751559153 Chr.16: 694588-694591 on Build GRCh38
rs753909394 Chr.16: 694622-694623 on Build GRCh38
rs758015571 Chr.16: 694596-694596 on Build GRCh38
rs528394512 Chr.16: 694570-694570 on Build GRCh38
rs759977765 Chr.16: 694590-694590 on Build GRCh38
rs944402067 Chr.16: 694712-694712 on Build GRCh38
rs752396410 Chr.16: 694591-694591 on Build GRCh38
rs930521845 Chr.16: 694731-694731 on Build GRCh38
rs764392612 Chr.16: 694749-694749 on Build GRCh38
rs564177813 Chr.16: 694560-694560 on Build GRCh38
rs761147545 Chr.16: 694584-694584 on Build GRCh38
rs990828753 Chr.16: 694585-694586 on Build GRCh38
rs1005000815 Chr.16: 694578-694578 on Build GRCh38
rs747152872 Chr.16: 694727-694727 on Build GRCh38
rs1005443398 Chr.16: 694609-694609 on Build GRCh38
rs763245095 Chr.16: 694661-694661 on Build GRCh38
rs776542744 Chr.16: 694735-694735 on Build GRCh38
rs775909080 Chr.16: 694743-694743 on Build GRCh38
rs777748773 Chr.16: 694623-694623 on Build GRCh38
rs756811787 Chr.16: 694682-694682 on Build GRCh38
rs763738253 Chr.16: 694662-694662 on Build GRCh38
rs747119837 Chr.16: 694628-694628 on Build GRCh38
rs778175775 Chr.16: 694724-694724 on Build GRCh38
rs745741161 Chr.16: 694604-694604 on Build GRCh38
rs751083095 Chr.16: 694599-694599 on Build GRCh38
rs1023971226 Chr.16: 694680-694680 on Build GRCh38
rs374983732 Chr.16: 694585-694585 on Build GRCh38
rs763338527 Chr.16: 694745-694745 on Build GRCh38
rs777218849 Chr.16: 694640-694640 on Build GRCh38
rs754755821 Chr.16: 694703-694703 on Build GRCh38
rs754235829 Chr.16: 694589-694589 on Build GRCh38
rs568266500 Chr.16: 694638-694638 on Build GRCh38
rs1038663640 Chr.16: 694715-694715 on Build GRCh38
rs377317985 Chr.16: 694740-694740 on Build GRCh38
rs754900841 Chr.16: 694622-694622 on Build GRCh38
rs775818903 Chr.16: 694651-694651 on Build GRCh38
rs528931889 Chr.16: 694744-694744 on Build GRCh38
rs749112852 Chr.16: 694702-694702 on Build GRCh38
rs368681212 Chr.16: 694623-694623 on Build GRCh38
rs771094311 Chr.16: 694730-694730 on Build GRCh38
rs17855603 Chr.16: 694639-694639 on Build GRCh38
rs1012611448 Chr.16: 694665-694665 on Build GRCh38
rs866963270 Chr.16: 694652-694652 on Build GRCh38
rs780055996 Chr.16: 694688-694688 on Build GRCh38
rs759937987 Chr.16: 694649-694649 on Build GRCh38
rs971564226 Chr.16: 694717-694717 on Build GRCh38
rs766911599 Chr.16: 694683-694683 on Build GRCh38
rs950796166 Chr.16: 694573-694573 on Build GRCh38
rs765630955 Chr.16: 694650-694650 on Build GRCh38
rs368373246 Chr.16: 694627-694627 on Build GRCh38
rs781219083 Chr.16: 694601-694601 on Build GRCh38
rs779959884 Chr.16: 694613-694613 on Build GRCh38
rs916828009 Chr.16: 694595-694595 on Build GRCh38
rs374470559 Chr.16: 694625-694625 on Build GRCh38
rs751131152 Chr.16: 694670-694670 on Build GRCh38
rs894171479 Chr.16: 694660-694660 on Build GRCh38

View Full Product Details