Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70216862-70217123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A1
Entrez Gene ID
1297
Gene Name
collagen type IX alpha 1
Gene Aliases
DJ149L1.1.2, EDM6, MED, STL4
UniGene
Hs.590892
Gene Chromosome Location
Chr.6: 70216040-70303083 on Build GRCh38
Transcripts
NM_001851.4 NM_078485.3 XM_011535429.2 XM_011535430.2 XM_017010246.1 XM_017010247.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5774968 c.2742A>T p.A914A Chr.6: 70216921-70216921 on Build GRCh38
1195521 c.1981G>A p.A661T Chr.6: 70216953-70216953 on Build GRCh38
6471369 c.2036G>A p.*679* Chr.6: 70216898-70216898 on Build GRCh38
4483355 c.2694C>T p.P898P Chr.6: 70216969-70216969 on Build GRCh38
3629316 c.2630G>A p.R877Q Chr.6: 70217033-70217033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969476871 Chr.6: 70217076-70217076 on Build GRCh38
rs767536535 Chr.6: 70216986-70216986 on Build GRCh38
rs760617713 Chr.6: 70216988-70216988 on Build GRCh38
rs745421286 Chr.6: 70217073-70217073 on Build GRCh38
rs567606245 Chr.6: 70216942-70216942 on Build GRCh38
rs374561398 Chr.6: 70216902-70216902 on Build GRCh38
rs199581593 Chr.6: 70217040-70217040 on Build GRCh38
rs1056921 Chr.6: 70217054-70217054 on Build GRCh38
rs1031413732 Chr.6: 70217020-70217020 on Build GRCh38
rs747451078 Chr.6: 70216935-70216935 on Build GRCh38
rs772906014 Chr.6: 70216938-70216938 on Build GRCh38
rs149139869 Chr.6: 70217041-70217041 on Build GRCh38
rs981306729 Chr.6: 70217077-70217077 on Build GRCh38
rs145221543 Chr.6: 70216926-70216926 on Build GRCh38
rs376876121 Chr.6: 70216936-70216936 on Build GRCh38
rs758388716 Chr.6: 70217034-70217034 on Build GRCh38
rs202176764 Chr.6: 70217078-70217078 on Build GRCh38
rs898990280 Chr.6: 70216912-70216912 on Build GRCh38
rs770502831 Chr.6: 70216968-70216968 on Build GRCh38
rs763970167 Chr.6: 70216992-70216992 on Build GRCh38
rs370064762 Chr.6: 70216910-70216910 on Build GRCh38
rs370324739 Chr.6: 70216969-70216969 on Build GRCh38
rs764914222 Chr.6: 70216956-70216956 on Build GRCh38
rs957580783 Chr.6: 70216985-70216985 on Build GRCh38
rs201520437 Chr.6: 70217045-70217045 on Build GRCh38
rs773848465 Chr.6: 70217050-70217050 on Build GRCh38
rs879430201 Chr.6: 70216925-70216925 on Build GRCh38
rs773034538 Chr.6: 70217087-70217088 on Build GRCh38
rs1056920 Chr.6: 70217056-70217056 on Build GRCh38
rs764060012 Chr.6: 70217096-70217096 on Build GRCh38
rs775559838 Chr.6: 70217083-70217083 on Build GRCh38
rs778177942 Chr.6: 70216885-70216885 on Build GRCh38
rs745872166 Chr.6: 70216922-70216922 on Build GRCh38
rs758916884 Chr.6: 70216979-70216979 on Build GRCh38
rs569046385 Chr.6: 70217075-70217075 on Build GRCh38
rs886138293 Chr.6: 70216901-70216901 on Build GRCh38
rs1056923 Chr.6: 70217019-70217019 on Build GRCh38
rs1028844209 Chr.6: 70216952-70216952 on Build GRCh38
rs200829297 Chr.6: 70217046-70217046 on Build GRCh38
rs773938837 Chr.6: 70216974-70216974 on Build GRCh38
rs757561926 Chr.6: 70216994-70216994 on Build GRCh38
rs750538159 Chr.6: 70217026-70217026 on Build GRCh38
rs373430094 Chr.6: 70216909-70216909 on Build GRCh38
rs760704462 Chr.6: 70217084-70217084 on Build GRCh38
rs199845159 Chr.6: 70216987-70216987 on Build GRCh38

View Full Product Details