Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 21750635-21750878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFRA2
Entrez Gene ID
2675
Gene Name
GDNF family receptor alpha 2
Gene Aliases
GDNFRB, NRTNR-ALPHA, NTNRA, RETL2, TRNR2
UniGene
Hs.441202
Gene Chromosome Location
Chr.8: 21690403-21789296 on Build GRCh38
Transcripts
NM_001165038.1 NM_001165039.1 NM_001495.4 XM_006716327.3 XM_011544484.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4686829 c.744G>T p.K248N Chr.8: 21750638-21750638 on Build GRCh38
4686830 c.639C>T p.S213S Chr.8: 21750743-21750743 on Build GRCh38
6044160 c.701G>A p.R234Q Chr.8: 21750681-21750681 on Build GRCh38
6180385 c.613C>A p.R205S Chr.8: 21750769-21750769 on Build GRCh38
5915785 c.682G>A p.A228T Chr.8: 21750700-21750700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973981150 Chr.8: 21750671-21750671 on Build GRCh38
rs774492736 Chr.8: 21750824-21750824 on Build GRCh38
rs749993949 Chr.8: 21750704-21750704 on Build GRCh38
rs770732315 Chr.8: 21750822-21750822 on Build GRCh38
rs979031547 Chr.8: 21750809-21750809 on Build GRCh38
rs778196839 Chr.8: 21750695-21750695 on Build GRCh38
rs944832144 Chr.8: 21750769-21750769 on Build GRCh38
rs781042387 Chr.8: 21750740-21750740 on Build GRCh38
rs565122102 Chr.8: 21750680-21750680 on Build GRCh38
rs769908261 Chr.8: 21750748-21750748 on Build GRCh38
rs375577535 Chr.8: 21750821-21750821 on Build GRCh38
rs774246701 Chr.8: 21750758-21750758 on Build GRCh38
rs372113145 Chr.8: 21750721-21750721 on Build GRCh38
rs368858488 Chr.8: 21750716-21750716 on Build GRCh38
rs777905177 Chr.8: 21750820-21750820 on Build GRCh38
rs754668539 Chr.8: 21750805-21750805 on Build GRCh38
rs767947492 Chr.8: 21750682-21750682 on Build GRCh38
rs536103646 Chr.8: 21750812-21750812 on Build GRCh38
rs756617946 Chr.8: 21750689-21750689 on Build GRCh38
rs751773049 Chr.8: 21750675-21750675 on Build GRCh38
rs765940919 Chr.8: 21750800-21750800 on Build GRCh38
rs745879940 Chr.8: 21750836-21750836 on Build GRCh38
rs532186500 Chr.8: 21750753-21750753 on Build GRCh38
rs767569598 Chr.8: 21750767-21750767 on Build GRCh38
rs775845928 Chr.8: 21750768-21750768 on Build GRCh38
rs143474711 Chr.8: 21750806-21750806 on Build GRCh38
rs764654147 Chr.8: 21750788-21750788 on Build GRCh38
rs754314934 Chr.8: 21750791-21750791 on Build GRCh38
rs750997748 Chr.8: 21750801-21750801 on Build GRCh38
rs199623540 Chr.8: 21750757-21750757 on Build GRCh38
rs757897910 Chr.8: 21750793-21750793 on Build GRCh38
rs1007800186 Chr.8: 21750741-21750741 on Build GRCh38
rs374570518 Chr.8: 21750795-21750805 on Build GRCh38
rs753020122 Chr.8: 21750687-21750687 on Build GRCh38
rs759832571 Chr.8: 21750761-21750761 on Build GRCh38
rs766573343 Chr.8: 21750667-21750667 on Build GRCh38
rs754540853 Chr.8: 21750729-21750729 on Build GRCh38
rs530960209 Chr.8: 21750743-21750743 on Build GRCh38
rs796065050 Chr.8: 21750657-21750657 on Build GRCh38
rs755938834 Chr.8: 21750813-21750813 on Build GRCh38
rs761015721 Chr.8: 21750775-21750775 on Build GRCh38
rs757924856 Chr.8: 21750715-21750715 on Build GRCh38
rs1044987238 Chr.8: 21750853-21750853 on Build GRCh38
rs749417514 Chr.8: 21750756-21750756 on Build GRCh38

View Full Product Details