Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48836052-48836325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX23
Entrez Gene ID
9416
Gene Name
DEAD-box helicase 23
Gene Aliases
PRPF28, SNRNP100, U5-100K, U5-100KD, prp28
UniGene
Hs.130098
Gene Chromosome Location
Chr.12: 48829756-48852174 on Build GRCh38
Transcripts
NM_004818.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
109705 c.1333T>C p.F445L Chr.12: 48836170-48836170 on Build GRCh38
3931639 c.1312G>A p.G438S Chr.12: 48836191-48836191 on Build GRCh38
223544 c.1351G>A p.V451I Chr.12: 48836152-48836152 on Build GRCh38
6573400 c.1293C>T p.I431I Chr.12: 48836210-48836210 on Build GRCh38
6641636 c.1256G>A p.R419H Chr.12: 48836247-48836247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372986765 Chr.12: 48836227-48836227 on Build GRCh38
rs758259209 Chr.12: 48836079-48836079 on Build GRCh38
rs746881934 Chr.12: 48836144-48836144 on Build GRCh38
rs748436278 Chr.12: 48836203-48836203 on Build GRCh38
rs376175660 Chr.12: 48836239-48836239 on Build GRCh38
rs774271044 Chr.12: 48836086-48836086 on Build GRCh38
rs778994458 Chr.12: 48836187-48836187 on Build GRCh38
rs762704880 Chr.12: 48836296-48836296 on Build GRCh38
rs757162328 Chr.12: 48836150-48836150 on Build GRCh38
rs773581583 Chr.12: 48836233-48836233 on Build GRCh38
rs761752687 Chr.12: 48836217-48836217 on Build GRCh38
rs764864417 Chr.12: 48836284-48836284 on Build GRCh38
rs768832955 Chr.12: 48836183-48836183 on Build GRCh38
rs763929446 Chr.12: 48836304-48836304 on Build GRCh38
rs1010308258 Chr.12: 48836286-48836286 on Build GRCh38
rs749299522 Chr.12: 48836180-48836180 on Build GRCh38
rs142861821 Chr.12: 48836228-48836228 on Build GRCh38
rs139638878 Chr.12: 48836155-48836155 on Build GRCh38
rs752327389 Chr.12: 48836288-48836288 on Build GRCh38
rs758955952 Chr.12: 48836270-48836270 on Build GRCh38
rs917824245 Chr.12: 48836247-48836247 on Build GRCh38
rs1057908 Chr.12: 48836252-48836252 on Build GRCh38
rs139969977 Chr.12: 48836170-48836170 on Build GRCh38
rs747828927 Chr.12: 48836280-48836283 on Build GRCh38

View Full Product Details