Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100600501-100600740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2L3
Entrez Gene ID
283431
Gene Name
growth arrest specific 2 like 3
Gene Aliases
G2L3
UniGene
Hs.20575
Gene Chromosome Location
Chr.12: 100573661-100628288 on Build GRCh38
Transcripts
NM_001303130.1 NM_001303131.1 NM_174942.2 XM_005268804.3 XM_011538219.2 XM_011538220.2 XM_011538221.2 XM_017019206.1

SNP Detail

SNP ID Chromosome Location
rs749291656 Chr.12: 100600542-100600542 on Build GRCh38
rs995174230 Chr.12: 100600617-100600617 on Build GRCh38
rs555481303 Chr.12: 100600646-100600646 on Build GRCh38
rs758461521 Chr.12: 100600543-100600543 on Build GRCh38
rs755988220 Chr.12: 100600531-100600531 on Build GRCh38
rs777692991 Chr.12: 100600539-100600539 on Build GRCh38
rs79713790 Chr.12: 100600687-100600687 on Build GRCh38
rs777140387 Chr.12: 100600644-100600644 on Build GRCh38
rs766987822 Chr.12: 100600634-100600634 on Build GRCh38
rs1036962089 Chr.12: 100600604-100600604 on Build GRCh38
rs770361336 Chr.12: 100600575-100600575 on Build GRCh38
rs959476298 Chr.12: 100600710-100600710 on Build GRCh38
rs768796285 Chr.12: 100600568-100600568 on Build GRCh38
rs780048838 Chr.12: 100600555-100600555 on Build GRCh38
rs183888273 Chr.12: 100600573-100600573 on Build GRCh38
rs368063453 Chr.12: 100600593-100600593 on Build GRCh38
rs752465157 Chr.12: 100600530-100600530 on Build GRCh38
rs1005450819 Chr.12: 100600610-100600610 on Build GRCh38
rs934339218 Chr.12: 100600566-100600566 on Build GRCh38
rs1015538721 Chr.12: 100600616-100600616 on Build GRCh38
rs534723727 Chr.12: 100600713-100600713 on Build GRCh38
rs763615758 Chr.12: 100600581-100600581 on Build GRCh38
rs759188766 Chr.12: 100600597-100600597 on Build GRCh38
rs1035057690 Chr.12: 100600689-100600689 on Build GRCh38
rs759885864 Chr.12: 100600656-100600656 on Build GRCh38
rs767311560 Chr.12: 100600525-100600525 on Build GRCh38
rs946974791 Chr.12: 100600529-100600529 on Build GRCh38
rs189499732 Chr.12: 100600583-100600583 on Build GRCh38
rs191501974 Chr.12: 100600570-100600570 on Build GRCh38
rs540110753 Chr.12: 100600567-100600567 on Build GRCh38
rs1048023887 Chr.12: 100600683-100600683 on Build GRCh38
rs773853969 Chr.12: 100600576-100600576 on Build GRCh38

View Full Product Details