Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50331615-50331888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM186A
Entrez Gene ID
121006
Gene Name
family with sequence similarity 186 member A
Gene Aliases
-
UniGene
Hs.133187
Gene Chromosome Location
Chr.12: 50326854-50396633 on Build GRCh38
Transcripts
NM_001145475.1 XM_006719231.3 XM_011537890.2 XM_011537892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
940327 c.165G>T p.K55N Chr.12: 50331678-50331678 on Build GRCh38
940325 c.167A>G p.K56R Chr.12: 50331676-50331676 on Build GRCh38
3739710 c.102A>G p.T34T Chr.12: 50331741-50331741 on Build GRCh38
4853419 c.6747C>T p.I2249I Chr.12: 50331771-50331771 on Build GRCh38
6412910 c.6722T>C p.L2241P Chr.12: 50331796-50331796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935422436 Chr.12: 50331841-50331841 on Build GRCh38
rs776469261 Chr.12: 50331854-50331854 on Build GRCh38
rs746328939 Chr.12: 50331803-50331803 on Build GRCh38
rs751858863 Chr.12: 50331756-50331756 on Build GRCh38
rs948020619 Chr.12: 50331833-50331833 on Build GRCh38
rs781282987 Chr.12: 50331781-50331781 on Build GRCh38
rs200446972 Chr.12: 50331646-50331646 on Build GRCh38
rs755062742 Chr.12: 50331825-50331826 on Build GRCh38
rs760710592 Chr.12: 50331648-50331648 on Build GRCh38
rs192789931 Chr.12: 50331837-50331837 on Build GRCh38
rs780601476 Chr.12: 50331852-50331852 on Build GRCh38
rs757256406 Chr.12: 50331771-50331771 on Build GRCh38
rs770729541 Chr.12: 50331806-50331806 on Build GRCh38
rs946907254 Chr.12: 50331855-50331855 on Build GRCh38
rs1052482058 Chr.12: 50331848-50331848 on Build GRCh38
rs893477781 Chr.12: 50331763-50331763 on Build GRCh38
rs962543491 Chr.12: 50331770-50331770 on Build GRCh38
rs1011198602 Chr.12: 50331778-50331778 on Build GRCh38
rs201742326 Chr.12: 50331732-50331732 on Build GRCh38
rs532380094 Chr.12: 50331804-50331804 on Build GRCh38
rs766219277 Chr.12: 50331652-50331652 on Build GRCh38
rs369232659 Chr.12: 50331739-50331739 on Build GRCh38
rs749598683 Chr.12: 50331858-50331858 on Build GRCh38
rs199775951 Chr.12: 50331734-50331734 on Build GRCh38
rs753691402 Chr.12: 50331674-50331674 on Build GRCh38
rs568054714 Chr.12: 50331826-50331826 on Build GRCh38
rs751628021 Chr.12: 50331709-50331709 on Build GRCh38
rs531889326 Chr.12: 50331683-50331683 on Build GRCh38
rs987251254 Chr.12: 50331835-50331835 on Build GRCh38
rs951143358 Chr.12: 50331689-50331689 on Build GRCh38

View Full Product Details