Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6165158-6165399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH3
Entrez Gene ID
9265
Gene Name
cytohesin 3
Gene Aliases
ARNO3, GRP1, PSCD3
UniGene
Hs.487479
Gene Chromosome Location
Chr.7: 6161779-6272611 on Build GRCh38
Transcripts
NM_004227.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1091138 c.1076G>T p.R359L Chr.7: 6165324-6165324 on Build GRCh38
4459356 c.866C>T p.S289F Chr.7: 6165279-6165279 on Build GRCh38
6639583 c.1031C>T p.T344I Chr.7: 6165369-6165369 on Build GRCh38
4676066 c.847_849delAAG p.K283delK Chr.7: 6165296-6165298 on Build GRCh38
4781570 c.872+3G>A p.? Chr.7: 6165270-6165270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867287441 Chr.7: 6165308-6165308 on Build GRCh38
rs199616562 Chr.7: 6165359-6165359 on Build GRCh38
rs1030842003 Chr.7: 6165266-6165266 on Build GRCh38
rs975201140 Chr.7: 6165187-6165187 on Build GRCh38
rs771504465 Chr.7: 6165356-6165356 on Build GRCh38
rs749584345 Chr.7: 6165326-6165326 on Build GRCh38
rs371624575 Chr.7: 6165324-6165324 on Build GRCh38
rs559043546 Chr.7: 6165240-6165240 on Build GRCh38
rs765997383 Chr.7: 6165362-6165362 on Build GRCh38
rs966043103 Chr.7: 6165186-6165186 on Build GRCh38
rs550985739 Chr.7: 6165215-6165215 on Build GRCh38
rs199632916 Chr.7: 6165227-6165227 on Build GRCh38
rs746044558 Chr.7: 6165312-6165312 on Build GRCh38
rs139638792 Chr.7: 6165305-6165305 on Build GRCh38
rs746246194 Chr.7: 6165265-6165265 on Build GRCh38
rs569610057 Chr.7: 6165216-6165216 on Build GRCh38
rs73675861 Chr.7: 6165244-6165244 on Build GRCh38
rs771227083 Chr.7: 6165259-6165259 on Build GRCh38
rs747038594 Chr.7: 6165258-6165258 on Build GRCh38
rs909637053 Chr.7: 6165212-6165212 on Build GRCh38
rs778048265 Chr.7: 6165311-6165311 on Build GRCh38
rs779983703 Chr.7: 6165253-6165253 on Build GRCh38
rs774705612 Chr.7: 6165341-6165341 on Build GRCh38
rs565324688 Chr.7: 6165180-6165180 on Build GRCh38
rs367892641 Chr.7: 6165246-6165246 on Build GRCh38
rs201472863 Chr.7: 6165278-6165278 on Build GRCh38
rs76764492 Chr.7: 6165238-6165238 on Build GRCh38
rs766173996 Chr.7: 6165257-6165257 on Build GRCh38
rs200855765 Chr.7: 6165232-6165232 on Build GRCh38
rs763619168 Chr.7: 6165242-6165242 on Build GRCh38
rs370170994 Chr.7: 6165256-6165256 on Build GRCh38
rs200832651 Chr.7: 6165250-6165250 on Build GRCh38
rs1054135898 Chr.7: 6165229-6165229 on Build GRCh38
rs762373616 Chr.7: 6165241-6165241 on Build GRCh38
rs371435369 Chr.7: 6165252-6165252 on Build GRCh38
rs374730483 Chr.7: 6165254-6165254 on Build GRCh38
rs144116589 Chr.7: 6165275-6165275 on Build GRCh38
rs369467659 Chr.7: 6165293-6165293 on Build GRCh38
rs759949684 Chr.7: 6165239-6165239 on Build GRCh38
rs373802531 Chr.7: 6165243-6165244 on Build GRCh38
rs146476912 Chr.7: 6165299-6165299 on Build GRCh38
rs2301910 Chr.7: 6165368-6165368 on Build GRCh38
rs921148803 Chr.7: 6165211-6165211 on Build GRCh38
rs1028537356 Chr.7: 6165199-6165199 on Build GRCh38
rs551111893 Chr.7: 6165369-6165369 on Build GRCh38
rs771222802 Chr.7: 6165237-6165237 on Build GRCh38
rs939308894 Chr.7: 6165219-6165219 on Build GRCh38
rs948442724 Chr.7: 6165325-6165325 on Build GRCh38
rs776991331 Chr.7: 6165263-6165263 on Build GRCh38
rs200058389 Chr.7: 6165247-6165247 on Build GRCh38
rs542536904 Chr.7: 6165298-6165298 on Build GRCh38
rs759112712 Chr.7: 6165296-6165298 on Build GRCh38
rs532766605 Chr.7: 6165188-6165188 on Build GRCh38
rs201932588 Chr.7: 6165228-6165228 on Build GRCh38
rs761402109 Chr.7: 6165353-6165353 on Build GRCh38
rs780912262 Chr.7: 6165226-6165226 on Build GRCh38
rs772883077 Chr.7: 6165358-6165358 on Build GRCh38
rs539932644 Chr.7: 6165306-6165306 on Build GRCh38
rs767216711 Chr.7: 6165301-6165301 on Build GRCh38
rs986703442 Chr.7: 6165217-6165217 on Build GRCh38
rs778687948 Chr.7: 6165307-6165307 on Build GRCh38
rs560515376 Chr.7: 6165302-6165302 on Build GRCh38
rs983839764 Chr.7: 6165211-6165212 on Build GRCh38
rs769271999 Chr.7: 6165335-6165335 on Build GRCh38
rs373994734 Chr.7: 6165233-6165233 on Build GRCh38
rs748288823 Chr.7: 6165230-6165230 on Build GRCh38

View Full Product Details