Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26193696-26193969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER4
Entrez Gene ID
378807
Gene Name
cation channel sperm associated 4
Gene Aliases
-
UniGene
Hs.123532
Gene Chromosome Location
Chr.1: 26189962-26202542 on Build GRCh38
Transcripts
NM_198137.1 XM_011541432.2 XM_011541433.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4400683 c.379A>C p.T127P Chr.1: 26193808-26193808 on Build GRCh38
6864543 c.390C>T p.D130D Chr.1: 26193819-26193819 on Build GRCh38
6125708 c.432G>T p.W144C Chr.1: 26193861-26193861 on Build GRCh38
907589 c.372G>A p.L124L Chr.1: 26193801-26193801 on Build GRCh38
3977541 c.459+1G>A p.? Chr.1: 26193889-26193889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372475621 Chr.1: 26193795-26193795 on Build GRCh38
rs557994261 Chr.1: 26193924-26193924 on Build GRCh38
rs747314881 Chr.1: 26193753-26193753 on Build GRCh38
rs745586481 Chr.1: 26193746-26193746 on Build GRCh38
rs776066190 Chr.1: 26193750-26193750 on Build GRCh38
rs374876192 Chr.1: 26193926-26193926 on Build GRCh38
rs543943257 Chr.1: 26193948-26193948 on Build GRCh38
rs1002649076 Chr.1: 26193890-26193890 on Build GRCh38
rs915701262 Chr.1: 26193844-26193844 on Build GRCh38
rs552734631 Chr.1: 26193743-26193743 on Build GRCh38
rs144202417 Chr.1: 26193804-26193804 on Build GRCh38
rs569659645 Chr.1: 26193794-26193794 on Build GRCh38
rs368414018 Chr.1: 26193782-26193782 on Build GRCh38
rs1026459388 Chr.1: 26193921-26193921 on Build GRCh38
rs76511895 Chr.1: 26193904-26193904 on Build GRCh38
rs148720324 Chr.1: 26193817-26193817 on Build GRCh38
rs151280406 Chr.1: 26193863-26193863 on Build GRCh38
rs748746380 Chr.1: 26193919-26193919 on Build GRCh38
rs1035088630 Chr.1: 26193902-26193902 on Build GRCh38
rs770406546 Chr.1: 26193763-26193764 on Build GRCh38
rs753850462 Chr.1: 26193830-26193830 on Build GRCh38
rs371655731 Chr.1: 26193909-26193909 on Build GRCh38
rs762961571 Chr.1: 26193925-26193925 on Build GRCh38
rs185314400 Chr.1: 26193825-26193825 on Build GRCh38
rs763964660 Chr.1: 26193828-26193828 on Build GRCh38
rs752500886 Chr.1: 26193816-26193816 on Build GRCh38
rs144958654 Chr.1: 26193723-26193723 on Build GRCh38
rs771841104 Chr.1: 26193749-26193749 on Build GRCh38
rs770196485 Chr.1: 26193923-26193923 on Build GRCh38
rs769300057 Chr.1: 26193758-26193758 on Build GRCh38
rs201113211 Chr.1: 26193933-26193933 on Build GRCh38
rs759925509 Chr.1: 26193812-26193812 on Build GRCh38
rs777147282 Chr.1: 26193761-26193761 on Build GRCh38
rs997511580 Chr.1: 26193932-26193932 on Build GRCh38
rs376226650 Chr.1: 26193737-26193737 on Build GRCh38
rs113968986 Chr.1: 26193785-26193785 on Build GRCh38
rs760966308 Chr.1: 26193721-26193721 on Build GRCh38
rs776831117 Chr.1: 26193916-26193916 on Build GRCh38
rs761832086 Chr.1: 26193769-26193769 on Build GRCh38
rs12138368 Chr.1: 26193801-26193801 on Build GRCh38
rs779796240 Chr.1: 26193891-26193891 on Build GRCh38
rs778545568 Chr.1: 26193861-26193861 on Build GRCh38
rs1018883672 Chr.1: 26193757-26193757 on Build GRCh38
rs757117408 Chr.1: 26193856-26193856 on Build GRCh38
rs758132329 Chr.1: 26193886-26193886 on Build GRCh38
rs773892215 Chr.1: 26193913-26193914 on Build GRCh38

View Full Product Details