Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61967099-61967356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTH1
Entrez Gene ID
2495
Gene Name
ferritin heavy chain 1
Gene Aliases
FHC, FTH, FTHL6, HFE5, PIG15, PLIF
UniGene
Hs.524910
Gene Chromosome Location
Chr.11: 61964285-61967660 on Build GRCh38
Transcripts
NM_002032.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
230231 c.86T>A p.L29H Chr.11: 61967340-61967340 on Build GRCh38
4746415 c.114G>75 p.E63fs*12 Chr.11: 61967312-61967312 on Build GRCh38
4632562 c.96C>T p.S32S Chr.11: 61967330-61967330 on Build GRCh38
224985 c.99C>A p.Y33* Chr.11: 61967327-61967327 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs533585798 Chr.11: 61967129-61967129 on Build GRCh38
rs933727166 Chr.11: 61967158-61967158 on Build GRCh38
rs747489814 Chr.11: 61967277-61967277 on Build GRCh38
rs1034952469 Chr.11: 61967215-61967215 on Build GRCh38
rs955382826 Chr.11: 61967236-61967236 on Build GRCh38
rs768136029 Chr.11: 61967300-61967300 on Build GRCh38
rs1014053969 Chr.11: 61967256-61967256 on Build GRCh38
rs932219966 Chr.11: 61967209-61967209 on Build GRCh38
rs1027626671 Chr.11: 61967318-61967318 on Build GRCh38
rs1052847425 Chr.11: 61967252-61967252 on Build GRCh38
rs920592121 Chr.11: 61967121-61967121 on Build GRCh38
rs944853561 Chr.11: 61967160-61967160 on Build GRCh38
rs904173668 Chr.11: 61967221-61967221 on Build GRCh38
rs1049448236 Chr.11: 61967212-61967212 on Build GRCh38
rs1002413403 Chr.11: 61967213-61967213 on Build GRCh38
rs1039345270 Chr.11: 61967141-61967141 on Build GRCh38
rs960776081 Chr.11: 61967259-61967259 on Build GRCh38
rs1000364569 Chr.11: 61967228-61967228 on Build GRCh38
rs1038516224 Chr.11: 61967203-61967203 on Build GRCh38
rs1005748632 Chr.11: 61967197-61967197 on Build GRCh38
rs866033742 Chr.11: 61967162-61967162 on Build GRCh38
rs528164427 Chr.11: 61967270-61967270 on Build GRCh38
rs559432387 Chr.11: 61967214-61967214 on Build GRCh38
rs776746010 Chr.11: 61967263-61967263 on Build GRCh38
rs748800078 Chr.11: 61967297-61967297 on Build GRCh38
rs983528688 Chr.11: 61967153-61967153 on Build GRCh38
rs887255173 Chr.11: 61967177-61967177 on Build GRCh38
rs779531508 Chr.11: 61967296-61967296 on Build GRCh38
rs778035914 Chr.11: 61967273-61967273 on Build GRCh38
rs908054160 Chr.11: 61967154-61967154 on Build GRCh38
rs771481657 Chr.11: 61967283-61967283 on Build GRCh38
rs1001387567 Chr.11: 61967278-61967278 on Build GRCh38
rs114039236 Chr.11: 61967163-61967163 on Build GRCh38
rs978566801 Chr.11: 61967301-61967301 on Build GRCh38
rs891624377 Chr.11: 61967253-61967253 on Build GRCh38
rs952056279 Chr.11: 61967321-61967321 on Build GRCh38
rs900790844 Chr.11: 61967200-61967200 on Build GRCh38
rs930751709 Chr.11: 61967148-61967148 on Build GRCh38
rs758825329 Chr.11: 61967271-61967271 on Build GRCh38

View Full Product Details