Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98001558-98001831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRR3
Entrez Gene ID
200959
Gene Name
gamma-aminobutyric acid type A receptor rho3 subunit (gene/pseudogene)
Gene Aliases
-
UniGene
Hs.534578
Gene Chromosome Location
Chr.3: 97986683-98035304 on Build GRCh38
Transcripts
NM_001105580.2 NR_047685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7212620 c.849G>T p.M283I Chr.3: 98001673-98001673 on Build GRCh38
1594099 c.849G>A p.M283I Chr.3: 98001673-98001673 on Build GRCh38
5732497 c.823T>C p.Y275H Chr.3: 98001699-98001699 on Build GRCh38
6011116 c.869G>A p.W290* Chr.3: 98001653-98001653 on Build GRCh38
1207805 c.781T>G p.F261V Chr.3: 98001741-98001741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753962830 Chr.3: 98001777-98001777 on Build GRCh38
rs143019522 Chr.3: 98001779-98001779 on Build GRCh38
rs564349175 Chr.3: 98001741-98001741 on Build GRCh38
rs766474256 Chr.3: 98001641-98001641 on Build GRCh38
rs745541183 Chr.3: 98001684-98001684 on Build GRCh38
rs372416866 Chr.3: 98001629-98001629 on Build GRCh38
rs759273883 Chr.3: 98001658-98001658 on Build GRCh38
rs377326407 Chr.3: 98001783-98001783 on Build GRCh38
rs772215382 Chr.3: 98001783-98001784 on Build GRCh38
rs770533657 Chr.3: 98001582-98001582 on Build GRCh38
rs762022068 Chr.3: 98001763-98001763 on Build GRCh38
rs779757579 Chr.3: 98001698-98001698 on Build GRCh38
rs561176393 Chr.3: 98001736-98001736 on Build GRCh38
rs80273016 Chr.3: 98001669-98001669 on Build GRCh38
rs760601957 Chr.3: 98001640-98001640 on Build GRCh38
rs751378563 Chr.3: 98001664-98001664 on Build GRCh38
rs998824184 Chr.3: 98001645-98001645 on Build GRCh38
rs760983133 Chr.3: 98001769-98001769 on Build GRCh38
rs749111895 Chr.3: 98001713-98001715 on Build GRCh38
rs769352971 Chr.3: 98001752-98001752 on Build GRCh38
rs750020706 Chr.3: 98001802-98001802 on Build GRCh38
rs1008957888 Chr.3: 98001624-98001624 on Build GRCh38
rs578242445 Chr.3: 98001646-98001646 on Build GRCh38
rs763575741 Chr.3: 98001609-98001609 on Build GRCh38
rs767273861 Chr.3: 98001766-98001766 on Build GRCh38
rs756937646 Chr.3: 98001796-98001796 on Build GRCh38
rs769119972 Chr.3: 98001618-98001618 on Build GRCh38
rs370299417 Chr.3: 98001712-98001712 on Build GRCh38
rs775704441 Chr.3: 98001806-98001806 on Build GRCh38
rs889223052 Chr.3: 98001615-98001615 on Build GRCh38
rs61730113 Chr.3: 98001695-98001695 on Build GRCh38
rs769360160 Chr.3: 98001605-98001605 on Build GRCh38
rs780750443 Chr.3: 98001600-98001600 on Build GRCh38
rs375460959 Chr.3: 98001804-98001804 on Build GRCh38
rs779706333 Chr.3: 98001807-98001807 on Build GRCh38
rs376249139 Chr.3: 98001743-98001743 on Build GRCh38
rs957278962 Chr.3: 98001725-98001725 on Build GRCh38
rs772487886 Chr.3: 98001764-98001764 on Build GRCh38
rs767687959 Chr.3: 98001635-98001635 on Build GRCh38
rs370915110 Chr.3: 98001800-98001800 on Build GRCh38
rs61730114 Chr.3: 98001726-98001726 on Build GRCh38
rs61733778 Chr.3: 98001759-98001759 on Build GRCh38
rs530681967 Chr.3: 98001639-98001639 on Build GRCh38
rs879175697 Chr.3: 98001688-98001688 on Build GRCh38
rs375712400 Chr.3: 98001644-98001644 on Build GRCh38
rs780653601 Chr.3: 98001680-98001680 on Build GRCh38
rs774839226 Chr.3: 98001753-98001753 on Build GRCh38
rs773009972 Chr.3: 98001601-98001601 on Build GRCh38
rs187334163 Chr.3: 98001634-98001634 on Build GRCh38
rs764077413 Chr.3: 98001782-98001782 on Build GRCh38
rs766426986 Chr.3: 98001771-98001771 on Build GRCh38
rs776074141 Chr.3: 98001608-98001608 on Build GRCh38
rs745429124 Chr.3: 98001603-98001603 on Build GRCh38

View Full Product Details