Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78111170-78111403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIB2
Entrez Gene ID
10518
Gene Name
calcium and integrin binding family member 2
Gene Aliases
DFNB48, KIP2, USH1J
UniGene
Hs.129867
Gene Chromosome Location
Chr.15: 78104606-78131976 on Build GRCh38
Transcripts
NM_001271888.1 NM_001271889.1 NM_001301224.1 NM_006383.3 NR_125435.1 XM_005254126.3 XM_006720374.2 XM_011521161.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1708423 c.102C>A p.F34L Chr.15: 78111261-78111261 on Build GRCh38
6860783 c.120C>A p.N40K Chr.15: 78111243-78111243 on Build GRCh38
701750 c.138C>A p.Y46* Chr.15: 78111225-78111225 on Build GRCh38
1940528 c.114C>T p.A38A Chr.15: 78111249-78111249 on Build GRCh38
1940530 c.95C>T p.S32L Chr.15: 78111268-78111268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1037264978 Chr.15: 78111337-78111337 on Build GRCh38
rs767412639 Chr.15: 78111325-78111325 on Build GRCh38
rs1003742354 Chr.15: 78111376-78111376 on Build GRCh38
rs774928839 Chr.15: 78111380-78111380 on Build GRCh38
rs560859905 Chr.15: 78111197-78111197 on Build GRCh38
rs760922217 Chr.15: 78111280-78111280 on Build GRCh38
rs769507153 Chr.15: 78111282-78111282 on Build GRCh38
rs779074778 Chr.15: 78111190-78111190 on Build GRCh38
rs199524336 Chr.15: 78111222-78111222 on Build GRCh38
rs367662751 Chr.15: 78111318-78111318 on Build GRCh38
rs751434719 Chr.15: 78111226-78111226 on Build GRCh38
rs886766317 Chr.15: 78111343-78111343 on Build GRCh38
rs769256040 Chr.15: 78111195-78111195 on Build GRCh38
rs550750703 Chr.15: 78111230-78111230 on Build GRCh38
rs757060895 Chr.15: 78111232-78111232 on Build GRCh38
rs751060652 Chr.15: 78111307-78111307 on Build GRCh38
rs117723991 Chr.15: 78111279-78111279 on Build GRCh38
rs767935017 Chr.15: 78111199-78111199 on Build GRCh38
rs151260624 Chr.15: 78111268-78111268 on Build GRCh38
rs749299160 Chr.15: 78111192-78111192 on Build GRCh38
rs142260703 Chr.15: 78111233-78111233 on Build GRCh38
rs900894840 Chr.15: 78111308-78111308 on Build GRCh38
rs750179982 Chr.15: 78111237-78111237 on Build GRCh38
rs756400008 Chr.15: 78111239-78111239 on Build GRCh38
rs764761715 Chr.15: 78111211-78111211 on Build GRCh38
rs755819035 Chr.15: 78111328-78111328 on Build GRCh38
rs201845656 Chr.15: 78111266-78111266 on Build GRCh38
rs199755684 Chr.15: 78111259-78111259 on Build GRCh38
rs149227237 Chr.15: 78111209-78111209 on Build GRCh38
rs888483463 Chr.15: 78111311-78111311 on Build GRCh38
rs541843030 Chr.15: 78111267-78111267 on Build GRCh38
rs749337651 Chr.15: 78111210-78111210 on Build GRCh38
rs774969149 Chr.15: 78111300-78111300 on Build GRCh38
rs371015361 Chr.15: 78111198-78111198 on Build GRCh38
rs75314194 Chr.15: 78111326-78111326 on Build GRCh38
rs897373243 Chr.15: 78111349-78111349 on Build GRCh38
rs1039513666 Chr.15: 78111284-78111284 on Build GRCh38
rs546124617 Chr.15: 78111336-78111336 on Build GRCh38
rs763540423 Chr.15: 78111305-78111305 on Build GRCh38
rs762318513 Chr.15: 78111217-78111217 on Build GRCh38
rs780117405 Chr.15: 78111240-78111240 on Build GRCh38
rs376449619 Chr.15: 78111301-78111301 on Build GRCh38
rs749410113 Chr.15: 78111245-78111245 on Build GRCh38
rs200314445 Chr.15: 78111216-78111216 on Build GRCh38
rs201062535 Chr.15: 78111203-78111203 on Build GRCh38

View Full Product Details