Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35631735-35632008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE1
Entrez Gene ID
8904
Gene Name
copine 1
Gene Aliases
COPN1, CPN1
UniGene
Hs.246413
Gene Chromosome Location
Chr.20: 35626031-35664956 on Build GRCh38
Transcripts
NM_001198863.1 NM_003915.5 NM_152925.2 NM_152926.2 NM_152927.2 NM_152928.2 NR_037188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3740103 c.588T>C p.R196R Chr.20: 35631742-35631742 on Build GRCh38
3701620 c.508T>A p.F170I Chr.20: 35631989-35631989 on Build GRCh38
1326779 c.515A>T p.Q172L Chr.20: 35631982-35631982 on Build GRCh38
4855409 c.550G>A p.E184K Chr.20: 35631947-35631947 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1046759758 Chr.20: 35631885-35631885 on Build GRCh38
rs763074393 Chr.20: 35631819-35631819 on Build GRCh38
rs773103707 Chr.20: 35631816-35631816 on Build GRCh38
rs111451577 Chr.20: 35631933-35631933 on Build GRCh38
rs757924953 Chr.20: 35631981-35631981 on Build GRCh38
rs563488892 Chr.20: 35631972-35631972 on Build GRCh38
rs761696013 Chr.20: 35631808-35631808 on Build GRCh38
rs745392563 Chr.20: 35631789-35631789 on Build GRCh38
rs2273356 Chr.20: 35631786-35631786 on Build GRCh38
rs910778740 Chr.20: 35631831-35631831 on Build GRCh38
rs897995698 Chr.20: 35631862-35631862 on Build GRCh38
rs781283297 Chr.20: 35631847-35631847 on Build GRCh38
rs370880449 Chr.20: 35631820-35631820 on Build GRCh38
rs907269561 Chr.20: 35631984-35631984 on Build GRCh38
rs771566331 Chr.20: 35631795-35631795 on Build GRCh38
rs145100771 Chr.20: 35631837-35631837 on Build GRCh38
rs753236790 Chr.20: 35631762-35631762 on Build GRCh38
rs143877309 Chr.20: 35631860-35631863 on Build GRCh38
rs1040629720 Chr.20: 35631845-35631845 on Build GRCh38
rs540101512 Chr.20: 35631880-35631880 on Build GRCh38
rs746707554 Chr.20: 35631799-35631799 on Build GRCh38
rs139024672 Chr.20: 35631930-35631930 on Build GRCh38
rs759724487 Chr.20: 35631905-35631905 on Build GRCh38
rs372765516 Chr.20: 35631950-35631950 on Build GRCh38
rs574803949 Chr.20: 35631861-35631861 on Build GRCh38
rs367911146 Chr.20: 35631809-35631809 on Build GRCh38
rs748102950 Chr.20: 35631823-35631824 on Build GRCh38
rs768410717 Chr.20: 35631803-35631803 on Build GRCh38
rs778437626 Chr.20: 35631936-35631936 on Build GRCh38
rs375119682 Chr.20: 35631827-35631827 on Build GRCh38
rs767946956 Chr.20: 35631916-35631916 on Build GRCh38
rs779743846 Chr.20: 35631796-35631796 on Build GRCh38
rs368111493 Chr.20: 35631863-35631866 on Build GRCh38
rs112341089 Chr.20: 35631942-35631942 on Build GRCh38
rs756714301 Chr.20: 35631766-35631766 on Build GRCh38
rs940695076 Chr.20: 35631848-35631848 on Build GRCh38
rs373786240 Chr.20: 35631806-35631806 on Build GRCh38
rs753144973 Chr.20: 35631919-35631919 on Build GRCh38
rs929483345 Chr.20: 35631878-35631878 on Build GRCh38
rs375741618 Chr.20: 35631985-35631985 on Build GRCh38

View Full Product Details