Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13798738-13798981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCUR1
Entrez Gene ID
63933
Gene Name
mitochondrial calcium uniporter regulator 1
Gene Aliases
C6orf79, CCDC90A, FMP32
UniGene
Hs.214043
Gene Chromosome Location
Chr.6: 13786549-13814994 on Build GRCh38
Transcripts
NM_001031713.3 XM_011514802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7311066 c.795C>T p.I265I Chr.6: 13798893-13798893 on Build GRCh38
5918876 c.791T>G p.V264G Chr.6: 13798897-13798897 on Build GRCh38
7310970 c.813C>T p.T271T Chr.6: 13798875-13798875 on Build GRCh38
3781978 c.855+4C>G p.? Chr.6: 13798829-13798829 on Build GRCh38
1620957 c.784-8C>A p.? Chr.6: 13798912-13798912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746142170 Chr.6: 13798856-13798856 on Build GRCh38
rs1008007572 Chr.6: 13798781-13798781 on Build GRCh38
rs764812955 Chr.6: 13798888-13798888 on Build GRCh38
rs769114151 Chr.6: 13798878-13798878 on Build GRCh38
rs764348234 Chr.6: 13798805-13798805 on Build GRCh38
rs757252606 Chr.6: 13798924-13798924 on Build GRCh38
rs990235930 Chr.6: 13798806-13798806 on Build GRCh38
rs746333062 Chr.6: 13798925-13798928 on Build GRCh38
rs781247235 Chr.6: 13798851-13798851 on Build GRCh38
rs371242312 Chr.6: 13798828-13798828 on Build GRCh38
rs768157416 Chr.6: 13798949-13798949 on Build GRCh38
rs75602422 Chr.6: 13798944-13798944 on Build GRCh38
rs781733737 Chr.6: 13798910-13798912 on Build GRCh38
rs757744637 Chr.6: 13798881-13798882 on Build GRCh38
rs778058404 Chr.6: 13798813-13798813 on Build GRCh38
rs77264261 Chr.6: 13798798-13798798 on Build GRCh38
rs879640550 Chr.6: 13798848-13798848 on Build GRCh38
rs375446121 Chr.6: 13798943-13798943 on Build GRCh38
rs757376760 Chr.6: 13798840-13798840 on Build GRCh38
rs765149010 Chr.6: 13798799-13798799 on Build GRCh38
rs372193345 Chr.6: 13798886-13798886 on Build GRCh38
rs762869084 Chr.6: 13798893-13798893 on Build GRCh38
rs747395967 Chr.6: 13798863-13798863 on Build GRCh38
rs7341222 Chr.6: 13798955-13798955 on Build GRCh38
rs977364778 Chr.6: 13798876-13798876 on Build GRCh38
rs760082802 Chr.6: 13798880-13798880 on Build GRCh38
rs145065022 Chr.6: 13798865-13798865 on Build GRCh38
rs751645323 Chr.6: 13798830-13798830 on Build GRCh38
rs914208730 Chr.6: 13798817-13798817 on Build GRCh38
rs368620336 Chr.6: 13798934-13798934 on Build GRCh38
rs375899276 Chr.6: 13798829-13798829 on Build GRCh38
rs151200565 Chr.6: 13798794-13798794 on Build GRCh38
rs752470539 Chr.6: 13798889-13798889 on Build GRCh38
rs971365583 Chr.6: 13798892-13798892 on Build GRCh38
rs751592313 Chr.6: 13798899-13798899 on Build GRCh38
rs778751432 Chr.6: 13798938-13798938 on Build GRCh38
rs767701745 Chr.6: 13798930-13798930 on Build GRCh38
rs764070202 Chr.6: 13798897-13798897 on Build GRCh38
rs367961059 Chr.6: 13798885-13798885 on Build GRCh38
rs770518075 Chr.6: 13798883-13798883 on Build GRCh38
rs879087714 Chr.6: 13798926-13798928 on Build GRCh38
rs770047913 Chr.6: 13798863-13798863 on Build GRCh38

View Full Product Details