Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45833095-45833342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRHR1
Entrez Gene ID
1394
Gene Name
corticotropin releasing hormone receptor 1
Gene Aliases
CRF-R, CRF-R-1, CRF-R1, CRF1, CRFR-1, CRFR1, CRH-R-1, CRH-R1, CRHR, CRHR1L
UniGene
Hs.417628
Gene Chromosome Location
Chr.17: 45784280-45835828 on Build GRCh38
Transcripts
NM_001145146.1 NM_001145147.1 NM_001145148.1 NM_001303018.1 NM_001303020.1 NM_004382.4 NM_001256299.2 NM_001303016.1

Gene Symbol
MGC57346-CRHR1
Entrez Gene ID
104909134
Gene Name
MGC57346-CRHR1 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.17: 45620329-45835828 on Build GRCh38
Transcripts
NM_001145146.1 NM_001145147.1 NM_001145148.1 NM_001303018.1 NM_001303020.1 NM_004382.4 NM_001256299.2 NM_001303016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4674677 c.809A>G p.Y270C Chr.17: 45833176-45833176 on Build GRCh38
7098665 c.773G>T p.C258F Chr.17: 45833140-45833140 on Build GRCh38
7098666 c.470G>T p.C157F Chr.17: 45833140-45833140 on Build GRCh38
6635451 c.896A>G p.Y299C Chr.17: 45833176-45833176 on Build GRCh38
6146898 c.876G>T p.R292S Chr.17: 45833156-45833156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779320726 Chr.17: 45833166-45833166 on Build GRCh38
rs752193765 Chr.17: 45833162-45833162 on Build GRCh38
rs374217021 Chr.17: 45833257-45833257 on Build GRCh38
rs775867466 Chr.17: 45833229-45833230 on Build GRCh38
rs1001945869 Chr.17: 45833189-45833189 on Build GRCh38
rs375302875 Chr.17: 45833207-45833207 on Build GRCh38
rs767999892 Chr.17: 45833118-45833118 on Build GRCh38
rs1035912096 Chr.17: 45833210-45833210 on Build GRCh38
rs769412850 Chr.17: 45833251-45833251 on Build GRCh38
rs1052162629 Chr.17: 45833320-45833320 on Build GRCh38
rs79608615 Chr.17: 45833171-45833171 on Build GRCh38
rs557675216 Chr.17: 45833310-45833310 on Build GRCh38
rs377543639 Chr.17: 45833221-45833221 on Build GRCh38
rs762497778 Chr.17: 45833115-45833115 on Build GRCh38
rs760799631 Chr.17: 45833236-45833238 on Build GRCh38
rs754047393 Chr.17: 45833161-45833161 on Build GRCh38
rs372074985 Chr.17: 45833237-45833237 on Build GRCh38
rs371351168 Chr.17: 45833235-45833235 on Build GRCh38
rs16940674 Chr.17: 45833141-45833141 on Build GRCh38
rs969750424 Chr.17: 45833271-45833271 on Build GRCh38
rs370128802 Chr.17: 45833201-45833201 on Build GRCh38
rs538607875 Chr.17: 45833134-45833134 on Build GRCh38
rs115157945 Chr.17: 45833272-45833272 on Build GRCh38
rs374351159 Chr.17: 45833199-45833199 on Build GRCh38
rs772322878 Chr.17: 45833225-45833225 on Build GRCh38
rs777782408 Chr.17: 45833184-45833184 on Build GRCh38
rs766671085 Chr.17: 45833150-45833150 on Build GRCh38
rs758616111 Chr.17: 45833174-45833174 on Build GRCh38
rs242950 Chr.17: 45833287-45833287 on Build GRCh38
rs966418923 Chr.17: 45833282-45833282 on Build GRCh38
rs577619488 Chr.17: 45833315-45833315 on Build GRCh38

View Full Product Details