Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 99850478-99850705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMSS1
Entrez Gene ID
84319
Gene Name
cms1 ribosomal small subunit homolog (yeast)
Gene Aliases
C3orf26
UniGene
Hs.581320
Gene Chromosome Location
Chr.3: 99817834-100178632 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Gene Symbol
FILIP1L
Entrez Gene ID
11259
Gene Name
filamin A interacting protein 1 like
Gene Aliases
DOC-1, DOC1, GIP130, GIP90
UniGene
Hs.104672
Gene Chromosome Location
Chr.3: 99833144-100114513 on Build GRCh38
Transcripts
NM_032359.3 NM_001042459.2 NM_001282793.1 NM_001282794.1 NM_014890.3 NM_182909.3 XM_006713486.2 XM_011512371.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4457677 c.1054C>T p.Q352* Chr.3: 99850622-99850622 on Build GRCh38
3598486 c.1034G>A p.R345Q Chr.3: 99850642-99850642 on Build GRCh38
6246412 c.1138C>A p.L380I Chr.3: 99850538-99850538 on Build GRCh38
1594134 c.1132C>T p.R378C Chr.3: 99850544-99850544 on Build GRCh38
2950974 c.1076G>A p.S359N Chr.3: 99850600-99850600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368008149 Chr.3: 99850661-99850661 on Build GRCh38
rs751008182 Chr.3: 99850563-99850563 on Build GRCh38
rs999556807 Chr.3: 99850521-99850521 on Build GRCh38
rs757820734 Chr.3: 99850572-99850572 on Build GRCh38
rs776559519 Chr.3: 99850526-99850526 on Build GRCh38
rs759335251 Chr.3: 99850682-99850682 on Build GRCh38
rs960816465 Chr.3: 99850653-99850653 on Build GRCh38
rs748174973 Chr.3: 99850671-99850671 on Build GRCh38
rs542026629 Chr.3: 99850643-99850643 on Build GRCh38
rs763542461 Chr.3: 99850548-99850548 on Build GRCh38
rs770961757 Chr.3: 99850675-99850675 on Build GRCh38
rs371654121 Chr.3: 99850629-99850629 on Build GRCh38
rs374961168 Chr.3: 99850642-99850642 on Build GRCh38
rs770746244 Chr.3: 99850501-99850501 on Build GRCh38
rs1002188910 Chr.3: 99850550-99850550 on Build GRCh38
rs958089345 Chr.3: 99850652-99850655 on Build GRCh38
rs750287688 Chr.3: 99850543-99850543 on Build GRCh38
rs374462158 Chr.3: 99850522-99850522 on Build GRCh38
rs201150738 Chr.3: 99850533-99850533 on Build GRCh38
rs776377840 Chr.3: 99850681-99850681 on Build GRCh38
rs745929745 Chr.3: 99850642-99850642 on Build GRCh38
rs752275867 Chr.3: 99850535-99850535 on Build GRCh38
rs781544113 Chr.3: 99850581-99850581 on Build GRCh38
rs756489358 Chr.3: 99850624-99850624 on Build GRCh38
rs764887333 Chr.3: 99850534-99850534 on Build GRCh38
rs768978938 Chr.3: 99850651-99850651 on Build GRCh38
rs187751369 Chr.3: 99850649-99850649 on Build GRCh38
rs750856389 Chr.3: 99850615-99850615 on Build GRCh38
rs368700289 Chr.3: 99850544-99850544 on Build GRCh38
rs769533562 Chr.3: 99850686-99850686 on Build GRCh38
rs1032378475 Chr.3: 99850613-99850613 on Build GRCh38

View Full Product Details