Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 79794549-79794796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX8
Entrez Gene ID
57332
Gene Name
chromobox 8
Gene Aliases
PC3, RC1
UniGene
Hs.387258
Gene Chromosome Location
Chr.17: 79794377-79797116 on Build GRCh38
Transcripts
NM_020649.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6008328 c.1139C>T p.T380M Chr.17: 79794666-79794666 on Build GRCh38
196271 c.1155delT p.F385fs*>5 Chr.17: 79794650-79794650 on Build GRCh38
1238436 c.1055G>C p.S352T Chr.17: 79794750-79794750 on Build GRCh38
279596 c.1166G>T p.R389I Chr.17: 79794639-79794639 on Build GRCh38
3820846 c.1161G>T p.E387D Chr.17: 79794644-79794644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757839842 Chr.17: 79794593-79794593 on Build GRCh38
rs750306972 Chr.17: 79794710-79794710 on Build GRCh38
rs902077838 Chr.17: 79794577-79794577 on Build GRCh38
rs746626590 Chr.17: 79794604-79794604 on Build GRCh38
rs1039507252 Chr.17: 79794580-79794580 on Build GRCh38
rs568531389 Chr.17: 79794656-79794656 on Build GRCh38
rs200864050 Chr.17: 79794606-79794606 on Build GRCh38
rs767656272 Chr.17: 79794662-79794662 on Build GRCh38
rs775603727 Chr.17: 79794669-79794669 on Build GRCh38
rs761806531 Chr.17: 79794694-79794694 on Build GRCh38
rs753655247 Chr.17: 79794689-79794689 on Build GRCh38
rs139495141 Chr.17: 79794665-79794665 on Build GRCh38
rs749671641 Chr.17: 79794746-79794746 on Build GRCh38
rs751216903 Chr.17: 79794728-79794728 on Build GRCh38
rs771362834 Chr.17: 79794757-79794757 on Build GRCh38
rs62000363 Chr.17: 79794735-79794735 on Build GRCh38
rs373611099 Chr.17: 79794627-79794627 on Build GRCh38
rs761026586 Chr.17: 79794698-79794698 on Build GRCh38
rs760918886 Chr.17: 79794686-79794686 on Build GRCh38
rs779579608 Chr.17: 79794720-79794720 on Build GRCh38
rs773601292 Chr.17: 79794622-79794622 on Build GRCh38
rs979967432 Chr.17: 79794750-79794750 on Build GRCh38
rs548721125 Chr.17: 79794584-79794584 on Build GRCh38
rs778062318 Chr.17: 79794744-79794744 on Build GRCh38
rs757927746 Chr.17: 79794713-79794713 on Build GRCh38
rs927208196 Chr.17: 79794768-79794768 on Build GRCh38
rs1042733227 Chr.17: 79794592-79794592 on Build GRCh38
rs763845874 Chr.17: 79794688-79794688 on Build GRCh38
rs778452438 Chr.17: 79794589-79794589 on Build GRCh38
rs200134960 Chr.17: 79794739-79794739 on Build GRCh38
rs753760461 Chr.17: 79794588-79794588 on Build GRCh38
rs771556709 Chr.17: 79794632-79794632 on Build GRCh38
rs774928845 Chr.17: 79794641-79794641 on Build GRCh38
rs368785850 Chr.17: 79794600-79794600 on Build GRCh38
rs917101305 Chr.17: 79794703-79794703 on Build GRCh38
rs996460192 Chr.17: 79794581-79794581 on Build GRCh38

View Full Product Details