Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18924793-18925056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX49
Entrez Gene ID
54555
Gene Name
DEAD-box helicase 49
Gene Aliases
R27090_2
UniGene
Hs.143187
Gene Chromosome Location
Chr.19: 18919675-18928633 on Build GRCh38
Transcripts
NM_019070.4 NR_033677.1 XM_011528083.1 XM_011528084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6641883 c.1009G>A p.G337S Chr.19: 18924961-18924961 on Build GRCh38
6641884 c.717G>A p.S239S Chr.19: 18924961-18924961 on Build GRCh38
4075784 c.729C>A p.P243P Chr.19: 18924973-18924973 on Build GRCh38
6641885 c.957G>A p.V319V Chr.19: 18924909-18924909 on Build GRCh38
4850744 c.1008C>T p.V336V Chr.19: 18924960-18924960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761435527 Chr.19: 18924831-18924831 on Build GRCh38
rs762773095 Chr.19: 18924814-18924814 on Build GRCh38
rs185451756 Chr.19: 18924834-18924834 on Build GRCh38
rs774001199 Chr.19: 18924816-18924816 on Build GRCh38
rs760140411 Chr.19: 18924945-18924945 on Build GRCh38
rs745333937 Chr.19: 18924854-18924855 on Build GRCh38
rs200828253 Chr.19: 18924839-18924839 on Build GRCh38
rs754375840 Chr.19: 18924845-18924845 on Build GRCh38
rs747807862 Chr.19: 18924927-18924927 on Build GRCh38
rs139952910 Chr.19: 18924931-18924931 on Build GRCh38
rs764360429 Chr.19: 18924971-18924971 on Build GRCh38
rs751859175 Chr.19: 18924976-18924976 on Build GRCh38
rs757336578 Chr.19: 18924993-18924993 on Build GRCh38
rs749182786 Chr.19: 18924904-18924904 on Build GRCh38
rs765629684 Chr.19: 18924968-18924968 on Build GRCh38
rs199956408 Chr.19: 18924874-18924874 on Build GRCh38
rs372637765 Chr.19: 18925025-18925025 on Build GRCh38
rs78976584 Chr.19: 18924969-18924969 on Build GRCh38
rs779158666 Chr.19: 18924857-18924857 on Build GRCh38
rs770238578 Chr.19: 18924870-18924870 on Build GRCh38
rs758669474 Chr.19: 18924970-18924970 on Build GRCh38
rs752949950 Chr.19: 18924862-18924862 on Build GRCh38
rs760019445 Chr.19: 18924960-18924960 on Build GRCh38
rs778749829 Chr.19: 18925019-18925019 on Build GRCh38
rs373999531 Chr.19: 18924899-18924899 on Build GRCh38
rs943885766 Chr.19: 18924897-18924897 on Build GRCh38
rs749558575 Chr.19: 18924955-18924955 on Build GRCh38
rs774085094 Chr.19: 18924919-18924919 on Build GRCh38
rs543975170 Chr.19: 18924930-18924930 on Build GRCh38
rs780097003 Chr.19: 18924900-18924900 on Build GRCh38
rs79331030 Chr.19: 18925002-18925002 on Build GRCh38
rs749247833 Chr.19: 18925001-18925001 on Build GRCh38
rs929034544 Chr.19: 18924974-18924974 on Build GRCh38
rs554978698 Chr.19: 18924855-18924855 on Build GRCh38
rs766997683 Chr.19: 18924833-18924833 on Build GRCh38
rs781431737 Chr.19: 18924997-18924997 on Build GRCh38
rs781017140 Chr.19: 18924847-18924847 on Build GRCh38
rs574885805 Chr.19: 18924865-18924865 on Build GRCh38
rs765878886 Chr.19: 18924948-18924948 on Build GRCh38
rs768489242 Chr.19: 18924917-18924917 on Build GRCh38

View Full Product Details