Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6974285-6974530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMG1
Entrez Gene ID
10436
Gene Name
EMG1, N1-specific pseudouridine methyltransferase
Gene Aliases
C2F, Grcc2f, NEP1
UniGene
Hs.558447
Gene Chromosome Location
Chr.12: 6970781-6997428 on Build GRCh38
Transcripts
NM_001320049.1 NM_006331.7 NR_135131.1

SNP Detail

SNP ID Chromosome Location
rs782521916 Chr.12: 6974386-6974386 on Build GRCh38
rs782735967 Chr.12: 6974421-6974421 on Build GRCh38
rs782703604 Chr.12: 6974438-6974438 on Build GRCh38
rs781896004 Chr.12: 6974432-6974432 on Build GRCh38
rs782241980 Chr.12: 6974504-6974504 on Build GRCh38
rs910414704 Chr.12: 6974350-6974350 on Build GRCh38
rs782419003 Chr.12: 6974318-6974318 on Build GRCh38
rs782174904 Chr.12: 6974364-6974364 on Build GRCh38
rs781815012 Chr.12: 6974374-6974374 on Build GRCh38
rs782634400 Chr.12: 6974311-6974311 on Build GRCh38
rs183111447 Chr.12: 6974322-6974322 on Build GRCh38
rs782531588 Chr.12: 6974461-6974461 on Build GRCh38
rs782532263 Chr.12: 6974467-6974467 on Build GRCh38
rs377272519 Chr.12: 6974385-6974385 on Build GRCh38
rs200644475 Chr.12: 6974317-6974317 on Build GRCh38
rs373473870 Chr.12: 6974353-6974353 on Build GRCh38
rs782446477 Chr.12: 6974402-6974403 on Build GRCh38
rs782115499 Chr.12: 6974356-6974356 on Build GRCh38
rs782509014 Chr.12: 6974309-6974309 on Build GRCh38
rs782803599 Chr.12: 6974430-6974430 on Build GRCh38
rs782601712 Chr.12: 6974400-6974400 on Build GRCh38
rs782600333 Chr.12: 6974503-6974503 on Build GRCh38
rs782485166 Chr.12: 6974452-6974452 on Build GRCh38
rs782793844 Chr.12: 6974357-6974357 on Build GRCh38
rs782693598 Chr.12: 6974403-6974403 on Build GRCh38
rs782707397 Chr.12: 6974371-6974371 on Build GRCh38
rs782339755 Chr.12: 6974352-6974352 on Build GRCh38
rs782179643 Chr.12: 6974402-6974402 on Build GRCh38
rs782172949 Chr.12: 6974389-6974389 on Build GRCh38
rs782689019 Chr.12: 6974387-6974387 on Build GRCh38
rs782016727 Chr.12: 6974359-6974359 on Build GRCh38
rs201857241 Chr.12: 6974334-6974334 on Build GRCh38
rs1053126796 Chr.12: 6974451-6974451 on Build GRCh38
rs782233120 Chr.12: 6974401-6974401 on Build GRCh38
rs370810704 Chr.12: 6974455-6974455 on Build GRCh38
rs74435397 Chr.12: 6974427-6974427 on Build GRCh38
rs781800072 Chr.12: 6974434-6974434 on Build GRCh38
rs782452445 Chr.12: 6974398-6974398 on Build GRCh38
rs782523942 Chr.12: 6974459-6974459 on Build GRCh38
rs782483867 Chr.12: 6974376-6974376 on Build GRCh38
rs782076226 Chr.12: 6974419-6974419 on Build GRCh38
rs782288416 Chr.12: 6974468-6974469 on Build GRCh38
rs782195719 Chr.12: 6974326-6974326 on Build GRCh38
rs782276276 Chr.12: 6974412-6974412 on Build GRCh38
rs781825544 Chr.12: 6974445-6974445 on Build GRCh38
rs782180735 Chr.12: 6974472-6974472 on Build GRCh38
rs782677923 Chr.12: 6974466-6974466 on Build GRCh38
rs782126371 Chr.12: 6974428-6974428 on Build GRCh38
rs782749715 Chr.12: 6974384-6974384 on Build GRCh38

View Full Product Details